Skip to Content
Merck
All Photos(4)

Key Documents

157260

Sigma-Aldrich

1-Hydroxybenzotriazole hydrate

98% dry basis wetted with not less than 14 wt. % water, for peptide synthesis

Synonym(s):

HOBt Hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3O · xH2O
CAS Number:
Molecular Weight:
135.12 (anhydrous basis)
Beilstein:
4515
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

1-Hydroxybenzotriazole hydrate, wetted with not less than 14 wt. % water, 98% dry basis

Quality Level

Assay

98% dry basis

form

solid

quality

synthesis grade

reaction suitability

reaction type: Addition Reactions

mp

155-158 °C (lit.)

application(s)

peptide synthesis

SMILES string

[H]O[H].On1nnc2ccccc12

InChI

1S/C6H5N3O.H2O/c10-9-6-4-2-1-3-5(6)7-8-9;/h1-4,10H;1H2

InChI key

PJUPKRYGDFTMTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hydroxybenzotriazole hydrate is a derivative of benzotriazole primarily used as a coupling reagent in peptide synthesis.[1][2][3]

Application

1-Hydroxybenzotriazole hydrate can be used as a reagent:
  • To synthesize β-peptides via microwave-assisted amino acid coupling.
  • In the spot synthesis of glycopeptides.
It can also be used as a deprotection agent for the removal of Fmoc (9-fluorenylmethoxycarbonyl) protecting group.

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Desen. Expl. 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 1

Flash Point(F)

314.6 °F

Flash Point(C)

157 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 35, 3315-3315 (1994)
Tetrahedron Letters, 34, 6383-6383 (1993)
T Katoh et al.
International journal of peptide and protein research, 42(3), 264-269 (1993-09-01)
3-Dimethylphosphinothioyl-2(3H)-oxazolone (MPTO) was synthesized, and its ability to effect racemization-free couplings and cyclization of a peptide and its C-terminal epimer was examined. MPTO showed good reactivity in aprotic polar solvents such as N,N-dimethylformamide (DMF) and N-methylpyrrolidone. In reactivity MPTO resembles
Synlett, 733-733 (1993)
Donna Wesolowski et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(40), 16582-16587 (2011-09-29)
Basic peptides covalently linked to nucleic acids, or chemically modified nucleic acids, enable the insertion of such a conjugate into bacteria grown in liquid medium and mammalian cells in tissue culture. A unique peptide, derived from human T cells, has

Questions

1–4 of 4 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Why is Product 157260, Hydroxybenzotriazole hydrate wet?

    1 answer
    1. Hydroxybenzotriazole hydrate is a solid that is explosive when dry. Our specification for this product is >14% water.

      Helpful?

  3. Can Product 157260, Hydroxybenzotriazole hydrate, be dried?

    1 answer
    1. Drying hydroxybenzotriazole hydrate should not be attempted because of its explosive nature.

      Helpful?

  4. How is Product 157260, Hydroxybenzotriazole hydrate, shipped?

    1 answer
    1. Hydroxybenzotriazole hydrate can not travel be sea or air. It is typically shipped by truck.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service