Skip to Content
Merck
All Photos(3)

Documents

151793

Sigma-Aldrich

Acetone-d6

99.9 atom % D

Synonym(s):

Hexadeuteroacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3COCD3
CAS Number:
Molecular Weight:
64.12
Beilstein:
1702935
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2 (vs air)

Quality Level

vapor pressure

14.39 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)

isotopic purity

99.9 atom % D

Assay

≥99% (CP)

form

liquid

expl. lim.

13.2 %

packaging

Sure/Seal of 100 g
ampule of 0.75 mL, 1 mL, 10 g
glass bottle of 25 g, 50 g

technique(s)

NMR: suitable

impurities

≤0.0500% water
water

refractive index

n20/D 1.355 (lit.)

bp

55.5 °C (lit.)

mp

−93.8 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]

InChI

1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3

InChI key

CSCPPACGZOOCGX-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone-d6 (CD3COCD3, Hexadeuteroacetone) is the hexadeuterated form of acetone, in which all hydrogen atoms in acetone molecule have been replaced by deuterium (D) atoms. It is a standard purity solvent for NMR (Nuclear Magnetic Resonance) analyses. Absorption spectra of CD3COCD3 samples cooled in molecular jet (jet-cooled) have been evaluated in the range of 51000-54000cm-1. Collision of its molecular ions with a perdeuterated surface (C20D41 SAu) was investigated by ion-surface collision spectroscopy. The Infrared and Raman spectra of acetone-d6 have been analyzed. It undergoes H-2H exchange reaction with acetone d0 to afford acetone d1 and d5.

Application

Acetone-d6 may be used as a deuterated solvent in the 1H NMR spectral studies of iodine containing radiopaque poly(methacrylate)copolymers. It may also be used as a source of deuterium atoms in the synthesis of labelled sterols.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability of radiopaque iodine-containing biomaterials
Aldenhoff.J.BY, et al.
Biomaterials, 23(3), 881-886 (2002)
The synthesis of labeled sterols
Kovganko.V.N and Ananich.K.S
Chemistry of Natural Compounds, 35(4), 369-393 (1999)
The (n0-3s) Rydberg state of acetone: absorption spectroscopy of jet-cooled acetone and acetone-d6.
Gaines GA, et al.
The Journal of Physical Chemistry, 92(10), 2762-2766 (1988)
Vibrational spectra and assignment of acetone, ααα acetone-d3 and acetone-d6.
Dellepiane G and Overend J.
Spectrochimica Acta , 22(4), 593-614 (1966)
Reactive ion-surface collisions: Application of ionized acetone-d6, DMSO-d6 and pyridine-d5 as probes for the characterization of self-assembled monolayer films on gold.
Kane TE, et al.
Org. Mass Spectrom., 28(12), 1665-1673 (1993)

Articles

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Use this reference table to find the coupling values and chemical shifts of our NMR (deuterated) solvents. Melting and boiling points, molecular weight, density, and CAS number are also listed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service