Skip to Content
Merck
All Photos(2)

Key Documents

147893

Sigma-Aldrich

α,α,α-Trichlorotoluene

98%

Synonym(s):

(Trichloromethyl)benzene, Benzotrichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CCl3
CAS Number:
Molecular Weight:
195.47
Beilstein:
508152
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.77 (vs air)

Quality Level

vapor pressure

0.2 mmHg ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

411 °F

refractive index

n20/D 1.557 (lit.)

bp

219-223 °C/760 mmHg (lit.)
90-91 °C/15 mmHg (lit.)

mp

−7.5-−7 °C (lit.)

solubility

alcohol: soluble
benzene: soluble
diethyl ether: soluble
organic solvents: soluble
water: insoluble

density

1.38 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)c1ccccc1

InChI

1S/C7H5Cl3/c8-7(9,10)6-4-2-1-3-5-6/h1-5H

InChI key

XEMRAKSQROQPBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

FT-IR and FT-Raman spectra of α,α,α-trichlorotoluene has been studied.

Application

α,α,α-Trichlorotoluene was used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Martí et al.
Journal of chromatography. A, 1077(1), 68-73 (2005-07-02)
A study of the occurrence of chlorinated toluenes in Llobregat river (NE Spain) has been carried out. These compounds are currently being used in local textile industries as dye carriers and have replaced the common trichlorobenzene mixtures. Closed loop stripping
Benzotrichloride.
Report on carcinogens : carcinogen profiles, 12, 66-67 (2011-08-19)
A mortality study of workers in a factory manufacturing chlorinated toluenes.
T Sorahan et al.
The Annals of occupational hygiene, 27(2), 173-182 (1983-01-01)
O Wong
American journal of industrial medicine, 14(4), 417-431 (1988-01-01)
The cohort of this historical prospective mortality study consisted of 697 male employees at a chlorination plant. A majority of the cohort was potentially exposed to benzotrichloride, benzyl chloride, benzoyl chloride, and other related chemicals. The mortality experience of the
Tamar Kohn et al.
Environmental science & technology, 40(13), 4253-4260 (2006-07-22)
Cross-correlations of rate constants between a system of interest and a better-defined one have become popular as a tool in studying transformations of organic pollutants. A slope of unity (if the correlation is conducted on a log-log basis) in such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service