Skip to Content
Merck
All Photos(2)

Key Documents

139912

Sigma-Aldrich

Phytol

95%, mixture of isomers

Synonym(s):

3,7,11,15-Tetramethyl-2-hexadecen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3[CH(CH3)(CH2)3]3C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
296.53
Beilstein:
1726098
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

(Liquid or Viscous Liquid)

refractive index

n20/D 1.463 (lit.)

bp

202-204 °C/10 mmHg (lit.)

density

0.85 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)CCCC(C)CCCC(C)CCC\C(C)=C\CO

InChI

1S/C20H40O/c1-17(2)9-6-10-18(3)11-7-12-19(4)13-8-14-20(5)15-16-21/h15,17-19,21H,6-14,16H2,1-5H3/b20-15+

InChI key

BOTWFXYSPFMFNR-HMMYKYKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phytol is a diterpene alcohol and is widely used as a food additive and in medicinal field. It undergoes Mitsunobu reaction with 5-fluorouridine to yield the nucleoterpenes.

Application

Phytol was used to investigate the electroantennogram response of males and females of the potato tuber moth, Phthorimaea operculella to various plant volatiles. It was used in the synthesis of pristanic acid (2,6,10,14-tetramethylpentadecanoic acid).

Biochem/physiol Actions

Phytol exhibits potential anti-Schistosomal properties. It induces sedative and anxiolytic-like effects in mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

368.6 °F

Flash Point(C)

187 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P P Van Veldhoven et al.
The Journal of biological chemistry, 266(36), 24676-24683 (1991-12-25)
Isoprenoid (branched) fatty acids such as pristanic acid can be degraded via beta-oxidation in peroxisomes. We synthesized 2-methylpalmitoyl-CoA as a model substrate in order to study the first step of the peroxisomal beta-oxidation of branched fatty acids, catalyzed by an
Edith Malecki et al.
Chemistry & biodiversity, 10(12), 2209-2220 (2013-12-12)
The cancerostatic 5-fluorouridine (5-FUrd; 1) was sequentially sugar-protected by introduction of a 2',3'-O-heptylidene ketal group (→2), followed by 5'-O-monomethoxytritylation (→3). This fully protected derivative was submitted to Mitsunobu reactions with either phytol ((Z and E)-isomer) or nerol ((Z)-isomer) to yield
Jéssica Pereira Costa et al.
Brain research, 1547, 34-42 (2013-12-18)
Phytol, a branched chain unsaturated alcohol, is particularly interesting because it is an isolated compound from essential oils of different medicinal plants. The aim of this study was to evaluate the anxiolytic-like effects of phytol in animal models to clarify
P D Das et al.
Journal of biosciences, 32(2), 339-349 (2007-04-17)
Electroantennograms (EAGs)were recorded from males and females of the potato tuber moth,Phthorimaea operculella in response to a broad range of plant volatile compounds belonging to diverse chemical classes.The responses to 27 compounds were evaluated,which indicated significant differences in EAGs between
Chiara Baccelli et al.
Journal of natural products, 70(6), 910-917 (2007-06-06)
The diterpenes previously isolated from the leaves of Croton zambesicus were tested to evaluate their vasorelaxant activity on the Wistar rat aorta. Their vasorelaxant effect was compared to a series of synthetic trachylobanes and related polycyclic compounds on KCl- or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service