Skip to Content
Merck
All Photos(3)

Key Documents

134821

Sigma-Aldrich

Boron trifluoride-methanol solution

50% w/w in methanol

Synonym(s):

BF3 methanol, BF3MeOH, Boron Trifluoride - Methanol Complex, Boron fluoride methanol, Boron trifluoride methanol, Boron trifluoride-methanol complex, boron fluoride methanol, borontrifluoride methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · MeOH
CAS Number:
Molecular Weight:
99.85
Beilstein:
3611499
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

19.5 mmHg ( 20 °C)

form

liquid

autoignition temp.

788 °F

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

49-53% ( by NaOH, titration)
50% w/w in methanol

technique(s)

gas chromatography (GC): suitable

bp

59 °C/4 mmHg

density

1.203 g/mL at 25 °C

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

CO.FB(F)F

InChI

1S/CH4O.BF3/c1-2;2-1(3)4/h2H,1H3;

InChI key

JBXYCUKPDAAYAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the GC analysis of ritalinic acid. Alternative to Cl2CHCO2H for synthesis of purine rich oligomers.
Lewis acid used in the preparation of geminal bis-peroxides from ketals and enol ethers with t-butyl hydroperoxide. Reagent used for the mild deacetylation of amines.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron trifluoride-methanol complex as a non-depurinating detritylating agent in DNA synthesis.
M J Mitchell et al.
Nucleic acids research, 18(17), 5321-5321 (1990-09-11)
Synthesis, 2215-2215 (2005)
Tetrahedron Letters, 44, 2301-2301 (2003)
T Vu-Duc et al.
Journal of pharmaceutical and biomedical analysis, 10(2-3), 187-191 (1992-02-01)
Ritalinic acid (RITA), the major metabolite of methylphenidate (Ritaline) is extracted with a solid-phase C8 column. Following elution, methylation of the carboxylic group is performed with boron trifluoride-methanol as a reagent. The methyl ester of RITA which structurally corresponds to
C Hue-Beauvais et al.
Journal of animal science, 93(4), 1641-1655 (2015-05-29)
Alterations to the metabolic endocrine environment during early life are crucial to mammary gland development. Among these environmental parameters, the initial nutritional event after birth is the consumption of milk, which represents the first maternal support provided to mammalian newborns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service