Skip to Content
Merck
All Photos(1)

Key Documents

130915

Sigma-Aldrich

2-Bromophenol

98%

Synonym(s):

o-Bromophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OH
CAS Number:
Molecular Weight:
173.01
Beilstein:
1905115
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.589 (lit.)

bp

195 °C (lit.)

mp

3-7 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)
1.6235 g/mL at 25 °C

functional group

bromo

SMILES string

Oc1ccccc1Br

InChI

1S/C6H5BrO/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

VADKRMSMGWJZCF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromophenol was used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabin-Lucian Suraru et al.
The Journal of organic chemistry, 79(1), 128-139 (2013-11-28)
Here we report a selective method for the core-extension of naphthalene diimide (NDI) with two annulated indole rings leading to carbazolo[2,3-b]carbazole diimides (CbDIs) with exclusive syn-connectivity based on a regioselective nucleophilic substitution reaction of Br4-NDI with arylamines, followed by palladium-catalyzed
Donna E Fennell et al.
Applied and environmental microbiology, 70(2), 1169-1175 (2004-02-10)
Terminal restriction fragment length polymorphism analysis of reverse-transcribed 16S rRNA during periods of community flux was used as a tool to delineate the roles of the members of a 2-bromophenol-degrading, sulfate-reducing consortium. Starved, washed cultures were amended with 2-bromophenol plus
Catherine S Evans et al.
Environmental science & technology, 39(7), 2128-2134 (2005-05-06)
The homogeneous, gas-phase oxidative thermal degradation of 2-bromophenol was studied in a 1 cm i.d., fused silica flow reactor at a concentration of 88 ppm, reaction time of 2.0 s, over a temperature range from 300 to 1000 degrees C.
Christian Eidamshaus et al.
Organic letters, 10(19), 4211-4214 (2008-08-30)
A one-pot synthesis of benzofurans which utilizes a palladium-catalyzed enolate arylation is described. The process demonstrates broad substrate scope and provides differentially substituted benzofurans in moderate to excellent yields. The utility of the method is further demonstrated by the synthesis
Miho Uchida et al.
Journal of hazardous materials, 101(3), 231-238 (2003-08-26)
2-Bromophenol was reacted in aqueous sodium hydroxide at 200-250 degrees C. The decomposition rate was remarkably faster at 250 degrees C than at 225 or 200 degrees C, and the percentage debromination reached almost 100% in 1M NaOH at 250

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service