128570
2-Hexanol
99%
Synonym(s):
(±)-2-Hexanol, Butyl methyl carbinol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3(CH2)3CH(OH)CH3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1718996
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.414 (lit.)
bp
136 °C (lit.)
density
0.814 g/mL at 20 °C
0.81 g/mL at 25 °C (lit.)
functional group
hydroxyl
SMILES string
CCCCC(C)O
InChI
1S/C6H14O/c1-3-4-5-6(2)7/h6-7H,3-5H2,1-2H3
InChI key
QNVRIHYSUZMSGM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2-Hexanol is one of the constituent of Porella arboris-vitae extracts which has been determined by solid phase microextraction, gas chromatography-mass spectrometry (SPME GC-MS).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Amit Kumar Tyagi et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 382927-382927 (2013-02-01)
The chemical composition of Porella arboris-vitae extracts was determined by solid phase microextraction, gas chromatography-mass spectrometry (SPME GC-MS), and 66 constituents were identified. The dominant compounds in methanol extract of P. arboris-vitae were β-caryophyllene (14.7%), α-gurjunene (10.9%), α-selinene (10.8%), β-elemene
Zhi-Xia Zheng et al.
Electrophoresis, 25(18-19), 3263-3269 (2004-10-09)
A novel chiral microemulsion, which involved the use of chiral alcohols as cosurfactants, was demonstrated for the enantiomeric separation of a number of pharmaceutical drugs in microemulsion electrokinetic chromatography (MEEKC). The chiral alcohols investigated were optically active 2-alkanols, with the
S J Crosbie et al.
Human & experimental toxicology, 16(3), 131-137 (1997-03-01)
1. The in vitro metabolism of n-hexane was studied in rat liver, lung, brain and skeletal muscle microsomes and in microsomes prepared from cell lines expressing human cytochrome P-450 2E1 or 2B6. The hydroxylated metabolites of n-hexane were quantified by
J K Alifimoff et al.
Anesthesiology, 66(1), 55-59 (1987-01-01)
The Meyer-Overton rule has been interpreted to mean that general anesthetics act at a nonpolar site, either in a lipid bilayer or a protein. Optical isomers, also called enantiomers, are pairs of compounds with the same molecular formula and functional
Kenji Gomi et al.
Plant molecular biology, 53(1-2), 189-199 (2004-02-06)
An expression profile of genes induced by non-pathogenic Alternaria alternata on rough lemon leaves was obtained by sequencing 500 subtractive PCR clones generated from mRNA of leaves inoculated with the fungus after subtraction with that of non-inoculated leaves. About 6%
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service