Skip to Content
Merck
All Photos(1)

Key Documents

128511

Sigma-Aldrich

2,2,4,4,6,8,8-Heptamethylnonane

98%

Synonym(s):

HMN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2C(CH3)2CH2C(CH3)3
CAS Number:
Molecular Weight:
226.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.9 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

240 °C (lit.)

density

0.793 g/mL at 25 °C (lit.)

SMILES string

CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C

InChI

1S/C16H34/c1-13(10-14(2,3)4)11-16(8,9)12-15(5,6)7/h13H,10-12H2,1-9H3

InChI key

VCLJODPNBNEBKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,4,4,6,8,8-Heptamethylnonane was used to study the effects of loading and aging pyrene in soils. It was also used to study its long term effect on the cell surface hydrophobicity (CSH) of a hexane-degrading Pseudomonas aeruginosa strain and a toluene-degrading Pseudomonas putida strain.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Somayeh Kazemzadeh et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 82(10), 2134-2147 (2020-12-03)
This study aimed to find biosurfactant producing and crude oil-degrading bacteria able to decontaminate crude oil from wastewater. The bacteria that were isolated from contaminated sites in an oil refinery plant in Isfahan, Iran, were identified by 16S rDNA sequencing
T Ikeda et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(11), 1271-1280 (1988-11-01)
1. Exposure of rats to 1% (w/w) of 2,2,4,4,6,8,8-heptamethylnonane in the diet for 2 weeks resulted in marked induction of liver peroxisome proliferation as judged from electron micrography, elevated activities of hepatic catalase (36%), cyanide-insensitive palmitoyl-CoA oxidase (10-fold), carnitine acetyl
J Harder et al.
Biodegradation, 11(1), 55-63 (2001-02-24)
Plant volatile organic compounds are a major carbon source in nature. We studied the degradability of these substances by anaerobic microorganisms in enrichment cultures with representative essential oils as organic substrates and nitrate as electron acceptor. Lemon and pine needle
Paulius Palaima et al.
Orphanet journal of rare diseases, 14(1), 197-197 (2019-08-16)
Charcot-Marie-Tooth (CMT) disease is the most common inherited neuromuscular disorder characterized by wide clinical, genetic and pathomechanistic heterogeneity. Recently, the gene encoding peripheral myelin protein 2 (PMP2) was identified as a novel cause for CMT neuropathy with three mutations that
Kathlyn M Kirkwood et al.
Biodegradation, 19(6), 785-794 (2008-02-27)
Mixed bacterial cultures aerobically transformed decalin (decahydronaphthalene) dissolved in an immiscible carrier phase (heptamethylnonane; HMN) in liquid medium. Conversion was enhanced in the presence of decane, a readily degraded n-alkane, and/or HMN. Four Rhodococcus spp. isolates purified from one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service