Skip to Content
Merck
All Photos(2)

Key Documents

127566

Sigma-Aldrich

Bis(diphenylphosphino)methane

97%

Synonym(s):

DPPM, Methanediylbis[diphenylphosphine, Tetraphenylmethylenediphosphine, DPM, Methylenebis(diphenylphosphine)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2PCH2P(C6H5)2
CAS Number:
Molecular Weight:
384.39
Beilstein:
758760
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

118-119 °C (lit.)

functional group

phosphine

SMILES string

C(P(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C25H22P2/c1-5-13-22(14-6-1)26(23-15-7-2-8-16-23)21-27(24-17-9-3-10-18-24)25-19-11-4-12-20-25/h1-20H,21H2

InChI key

XGCDBGRZEKYHNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert R A Freund et al.
Dalton transactions (Cambridge, England : 2003), 43(37), 13988-14000 (2014-08-15)
A series of allyl modified nickelalactone derivatives of the general formula [(L)nNi{CH2C(CH3)C(CH3)CH2COO}] was synthesized via ligand exchange reactions in order to investigate the influence of the neutral ligand L on the structure and stability of these complexes. While the square
Jun-Ichi Wachino et al.
mBio, 11(2) (2020-03-19)
Production of metallo-β-lactamases (MBLs), which hydrolyze carbapenems, is a cause of carbapenem resistance in Enterobacteriaceae Development of effective inhibitors for MBLs is one approach to restore carbapenem efficacy in carbapenem-resistant Enterobacteriaceae (CRE). We report here that sulfamoyl heteroarylcarboxylic acids (SHCs)
Somayeh Behrouz et al.
Molecular diversity, 23(1), 147-164 (2018-08-11)
The design, synthesis, antinociceptive and β-adrenoceptor blocking activities of several eugenyloxy propanol azole derivatives have been described. In this synthesis, the reaction of eugenol with epichlorohydrin provided adducts 3 and 4 which were N-alkylated by diverse azoles to obtain the
Birge D Özel Duygan et al.
Communications biology, 3(1), 379-379 (2020-07-17)
The study of complex microbial communities typically entails high-throughput sequencing and downstream bioinformatics analyses. Here we expand and accelerate microbiota analysis by enabling cell type diversity quantification from multidimensional flow cytometry data using a supervised machine learning algorithm of standard
Walinka van Tol et al.
Journal of inherited metabolic disease, 42(5), 984-992 (2019-04-02)
The congenital disorders of glycosylation (CDG) are inborn errors of metabolism with a great genetic heterogeneity. Most CDG are caused by defects in the N-glycan biosynthesis, leading to multisystem phenotypes. However, the occurrence of tissue-restricted clinical symptoms in the various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service