Skip to Content
Merck
All Photos(2)

Key Documents

127019

Sigma-Aldrich

N-Methylethylenediamine

95%

Synonym(s):

2-(Methylamino)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2NHCH3
CAS Number:
Molecular Weight:
74.12
Beilstein:
878141
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.4395 (lit.)

bp

114-117 °C (lit.)

density

0.85 g/mL at 20 °C (lit.)

SMILES string

CNCCN

InChI

1S/C3H10N2/c1-5-3-2-4/h5H,2-4H2,1H3

InChI key

KFIGICHILYTCJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methylethylenediamine undergoes condensation with pyridine-2-carboxaldehyde to form an imine. It reacts with isoquinoline-5-sulfonyl chloride to form N-(2-aminoethyl)-N-methyl-isoquinoline-5-sulfonamide.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Neil Vasdev et al.
Bioorganic & medicinal chemistry, 16(9), 5277-5284 (2008-03-19)
Protein kinase A (PKA) is an important signal transduction target for drug development because it influences critical cellular processes implicated in neuropsychiatric illnesses such as major depressive disorder. The goal of the present study was to develop the first imaging
Nan Shen et al.
Beilstein journal of nanotechnology, 8, 2032-2044 (2017-10-20)
α-Fe
Sandip Mukherjee et al.
Dalton transactions (Cambridge, England : 2003), 42(11), 4019-4030 (2013-01-24)
Three new copper-azido complexes [Cu(4)(N(3))(8)(L(1))(2)](n) (1), [Cu(4)(N(3))(6)(L(2))(2)(H(2)O)(2)] (2), and [Cu(4)(N(3))(6)(L(3))(2)](n) (3) [L(1) is the imine resulting from the condensation of pyridine-2-carboxaldehyde with N-methylethylenediamine, HL(2) and HL(3) are the condensation products of 2-hydroxy-3-methoxybenzaldehyde with N,N-diethylethylenediamine and N-ethylethylenediamine respectively] have been synthesized
Anna Urciuolo et al.
PloS one, 15(5), e0232081-e0232081 (2020-05-07)
The reproduction of reliable in vitro models of human skeletal muscle is made harder by the intrinsic 3D structural complexity of this tissue. Here we coupled engineered hydrogel with 3D structural cues and specific mechanical properties to derive human 3D
Leslie S Hamachi et al.
Chemical science, 10(26), 6539-6552 (2019-08-02)
We report a method to control the composition and microstructure of CdSe1-x S x

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service