Skip to Content
Merck
All Photos(3)

Documents

123242

Sigma-Aldrich

Anthraquinone-2-sulfonic acid sodium salt monohydrate

97%

Synonym(s):

9,10-Dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt monohydrate, Sodium anthraquinone-2-sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7NaO5S · H2O
CAS Number:
Molecular Weight:
328.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

impurities

4.0-6.0% water (Karl Fischer)

SMILES string

O.[Na+].[O-]S(=O)(=O)c1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C14H8O5S.Na.H2O/c15-13-9-3-1-2-4-10(9)14(16)12-7-8(20(17,18)19)5-6-11(12)13;;/h1-7H,(H,17,18,19);;1H2/q;+1;/p-1

InChI key

KHILLYASAGTPOI-UHFFFAOYSA-M

General description

Anthraquinone-2-sulfonic acid sodium salt monohydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate.

Anthraquinone-2-sulfonic acid sodium salt monohydrate can be used as a redox indicator to enhance the sensitivity and selectivity of the DNA biosensor.

Application

Anthraquinone-2-sulfonic acid sodium salt monohydrate was used to investigate the effects of reactant concentration and synthesis parameters on the electrical conductivity of polypyrrole coated polyethyleneterephthalate fabrics. It has been used as dopant in the preparation of polypyrrole films by electrochemical polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Electrical property and stability of electrochemically synthesized polypyrrole films.
Oh KW, et al.
Journal of Applied Polymer Science. Applied Polymer Symposium, 91(6), 3659-3666 (2004)
Bulletin of the Chemical Society of Japan, 67, 1538-1538 (1994)
Kyungha Baik et al.
Polymers, 11(6) (2019-06-27)
Smart textiles for wearable devices require flexibility and a lightweight, so in this study, a soft polypyrrole (PPy) electrode system was integrated into a piezoelectric polyvinylidenefluoride (PVDF) energy harvester. The PVDF energy harvester integrated with a PPy electrode had the
A regenerable screen-printed DNA biosensor based on acrylic microsphere--gold nanoparticle composite for genetically modified soybean determination
Ulianas A, et al.
Sensors and Actuators B, Chemical, 190, 694-701 (2014)
Effect of synthesis parameters on the electrical conductivity of polypyrrole-coated poly (ethylene terephthalate) fabrics.
Kaynak A and Rafael B.
Polymer International, 52(6), 1021-1026 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service