Skip to Content
Merck
All Photos(1)

Key Documents

120685

Sigma-Aldrich

N-Cyclohexylformamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHC6H11
CAS Number:
Molecular Weight:
127.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

bp

113 °C/700 mmHg (lit.)

mp

36-41 °C (lit.)

functional group

amide

SMILES string

O=CNC1CCCCC1

InChI

1S/C7H13NO/c9-6-8-7-4-2-1-3-5-7/h6-7H,1-5H2,(H,8,9)

InChI key

SWGXDLRCJNEEGZ-UHFFFAOYSA-N

General description

N-Cyclohexylformamide binds to the complex of horse liver alcohol dehydrogenase with NADH and is similar to the Michaelis complex for aldehyde reduction catalyzed by the enzyme[1]. It is formed during hydration of cyclohexyl isocyanide catalyzed by a novel enzyme isonitrile hydratase from Pseudomonas putida N19-2 [2].

Application

N-Cyclohexylformamide was used in the synthesis of 2-methylidene-1,3-selenazolidine derivatives[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selenium-containing heterocycles from isoselenocyanates: synthesis of 2-methylidene-1, 3-selenazolidine derivatives.
Sommen GL, et al.
Tetrahedron, 62(14), 3344-3354 (2006)
H Deng et al.
Biochemistry, 37(40), 14267-14278 (1998-10-07)
The binding of N-cyclohexylformamide (CXF) to the complex of horse liver alcohol dehydrogenase with NADH mimics that of the Michaelis complex for aldehyde reduction catalyzed by the enzyme. The Raman spectra of bound CXF and its 13C- and 15N-substituted derivatives
M Goda et al.
The Journal of biological chemistry, 276(26), 23480-23485 (2001-04-18)
Isonitrile containing an N triple bond C triple bond was degraded by microorganism sp. N19-2, which was isolated from soil through a 2-month acclimatization culture in the presence of this compound. The isonitrile-degrading microorganism was identified as Pseudomonas putida. The
S Ramaswamy et al.
Biochemistry, 36(12), 3522-3527 (1997-03-25)
Amides are analogs of aldehydes and potent inhibitors of liver alcohol dehydrogenases. They can be used for structural studies and for inhibiting the metabolism of alcohols that form toxic products. We studied N-alkyl amides that bind to the enzyme-NADH complex
Uncompetitive inhibitors of alcohol dehydrogenases.
B V Plapp et al.
Advances in experimental medicine and biology, 463, 295-303 (1999-06-03)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service