Skip to Content
Merck
All Photos(2)

Documents

113115

Sigma-Aldrich

5-Methoxy-1-tetralone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
Beilstein:
2047383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

160-162 °C/7 mmHg (lit.)

mp

87-91 °C (lit.)

SMILES string

COc1cccc2C(=O)CCCc12

InChI

1S/C11H12O2/c1-13-11-7-3-4-8-9(11)5-2-6-10(8)12/h3-4,7H,2,5-6H2,1H3

InChI key

BRCPWISABURVIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Methoxy-1-tetralone is used in the preparation of hydroxy ketone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service