Skip to Content
Merck
All Photos(3)

Key Documents

112151

Sigma-Aldrich

2,4-Dichloroaniline

99%

Synonym(s):

2,4-Dichlorophenylamine, 4-Chloro-2-chloroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
Beilstein:
386422
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050667
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

245 °C (lit.)

mp

59-62 °C (lit.)

SMILES string

Nc1ccc(Cl)cc1Cl

InChI

1S/C6H5Cl2N/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

KQCMTOWTPBNWDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichloroaniline is degraded by Delftia tsuruhatensis H1. 2,4-Dichloroaniline metabolite is detected in human urine sample by GC/MS and high performance liquid chromatograph.

Application

  • Phototransformation of 2,4-Dichloroaniline: Studies the phototransformation of 2,4-Dichloroaniline in freshwater environments, crucial for understanding its environmental degradation and implications for water purification technologies (Ucun et al., 2021).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kilemade et al.
Environmental toxicology, 16(3), 253-259 (2001-06-21)
The aim of this work was to investigate the use of the heat shock protein, HSP 70, as a sublethal measurement of ecotoxicity and to identify if the amount of HSP 70 synthesized is proportional to the chemical stress applied.
Michael Kilemade et al.
Aquatic toxicology (Amsterdam, Netherlands), 63(3), 207-219 (2003-04-25)
Interest in and concern for the quality of the environment has prompted a great deal of research into methods of measuring and assessing changes in it. One problem of major interest is that of increasing amounts of mutagenic/carcinogenic chemicals generated
C Causserand et al.
Water research, 39(8), 1594-1600 (2005-05-10)
This study evaluates the performance of two nanofiltration membranes in removing a herbicide: dichloroaniline. The membranes, one polyamide and one cellulose acetate, have a cut-off in the range 150-300 g/mol (manufacturers' data). The experiments were carried out with solutions of
Si-Houy Lao et al.
Phytochemistry, 63(6), 653-661 (2003-07-05)
The metabolic fate of [UL-14C]-3,4-dichloroaniline (DCA) was investigated in Arabidopsis root cultures and soybean plants over a 48 h period following treatment via the root media. DCA was rapidly taken up by both species and metabolised, predominantly to N-malonyl-DCA in
Phototransformation of 2,4-dichloroaniline in a surface freshwater environment: effects on microbial assemblages.
H M Hwang et al.
Bulletin of environmental contamination and toxicology, 60(1), 81-87 (1998-03-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service