Skip to Content
Merck
All Photos(1)

Key Documents

104256

Sigma-Aldrich

Ethyl p-toluenesulfonate

98%

Synonym(s):

Ethyl 4-methylbenzenesulfonate, Ethyl tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3C2H5
CAS Number:
Molecular Weight:
200.25
Beilstein:
611213
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

refractive index

n20/D 1.511 (lit.)

bp

158-162 °C/10 mmHg (lit.)

mp

29-33 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

functional group

tosylate

SMILES string

CCOS(=O)(=O)c1ccc(C)cc1

InChI

1S/C9H12O3S/c1-3-12-13(10,11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3

InChI key

VRZVPALEJCLXPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl p-toluenesulfonate was used in a study to develop a fast and accurate method for determination of residues of some common alkylating agents employed in drug synthesis by in situ derivatization–headspace–gas chromatography–mass spectrometry[1]. Ethyl p-toluenesulfonate was used to develop an extraction method for methyl and ethyl esters of various sulfonic acids in active pharmaceutical ingredients using solid-phase microextraction coupled to GC/MS in the selected ion monitoring mode[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

316.4 °F - closed cup

Flash Point(C)

158 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Alzaga et al.
Journal of pharmaceutical and biomedical analysis, 45(3), 472-479 (2007-08-28)
A simple, reliable and fast procedure for the simultaneous determination of residues of some common alkylating agents (AAs), such as mesylates, besylates, tosylates and sulfates, employed in drug synthesis, has been developed by in situ derivatization-headspace-gas chromatography-mass spectrometry. Pentafluorothiophenol is
Ivelisse Colón et al.
Journal of pharmaceutical and biomedical analysis, 39(3-4), 477-485 (2005-06-14)
The development, optimization and validation of an extraction method for methyl and ethyl esters of various sulfonic acids is presented. The extraction and determination of these esters in active pharmaceutical ingredients (APIs) was accomplished using solid-phase microextraction coupled to GC/MS
Olivier Barret et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 56(4), 586-591 (2015-02-24)
PET with selective adenosine 2A receptor (A2A) radiotracers can be used to study a variety of neurodegenerative and neuropsychiatric disorders in vivo and to support drug-discovery studies targeting A2A. The aim of this study was to describe the first in

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service