Merck
  • Derivation of Pluripotent Stem Cells with In Vivo Embryonic and Extraembryonic Potency.
Derivation of Pluripotent Stem Cells with In Vivo Embryonic and Extraembryonic Potency.
Cell (2017-04-08)
Yang Yang, Bei Liu, Jun Xu, Jinlin Wang, Jun Wu, Cheng Shi, Yaxing Xu, Jiebin Dong, Chengyan Wang, Weifeng Lai, Jialiang Zhu, Liang Xiong, Dicong Zhu, Xiang Li, Weifeng Yang, Takayoshi Yamauchi, Atsushi Sugawara, Zhongwei Li, Fangyuan Sun, Xiangyun Li, Chen Li, Aibin He, Yaqin Du, Ting Wang, Chaoran Zhao, Haibo Li, Xiaochun Chi, Hongquan Zhang, Yifang Liu, Cheng Li, Shuguang Duo, Ming Yin, Huan Shen, Juan Carlos Izpisua Belmonte, Hongkui Deng
ABSTRACT
Of all known cultured stem cell types, pluripotent stem cells (PSCs) sit atop the landscape of developmental potency and are characterized by their ability to generate all cell types of an adult organism. However, PSCs show limited contribution to the extraembryonic placental tissues in vivo. Here, we show that a chemical cocktail enables the derivation of stem cells with unique functional and molecular features from mice and humans, designated as extended pluripotent stem (EPS) cells, which are capable of chimerizing both embryonic and extraembryonic tissues. Notably, a single mouse EPS cell shows widespread chimeric contribution to both embryonic and extraembryonic lineages in vivo and permits generating single-EPS-cell-derived mice by tetraploid complementation. Furthermore, human EPS cells exhibit interspecies chimeric competency in mouse conceptuses. Our findings constitute a first step toward capturing pluripotent stem cells with extraembryonic developmental potentials in culture and open new avenues for basic and translational research. VIDEO ABSTRACT.
Sigma-Aldrich
Anti-KRT8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Sigma-Aldrich
(S)-(+)-Dimethindene maleate, ≥98% (HPLC)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit