Merck
  • Highly sensitive and specific protein detection via combined capillary isoelectric focusing and proximity ligation.
Highly sensitive and specific protein detection via combined capillary isoelectric focusing and proximity ligation.
Scientific reports (2017-05-06)
Narendra Padhan, Junhong Yan, Annegret Boge, Elaine Scrivener, Helgi Birgisson, Agata Zieba, Mats Gullberg, Masood Kamali-Moghaddam, Lena Claesson-Welsh, Ulf Landegren
ABSTRACT
Detection and quantification of proteins and their post-translational modifications are crucial to decipher functions of complex protein networks in cell biology and medicine. Capillary isoelectric focusing together with antibody-based detection can resolve and identify proteins and their isoforms with modest sample input. However, insufficient sensitivity prevents detection of proteins present at low concentrations and antibody cross-reactivity results in unspecific detection that cannot be distinguished from bona fide protein isoforms. By using DNA-conjugated antibodies enhanced signals can be obtained via rolling circle amplification (RCA). Both sensitivity and specificity can be greatly improved in assays dependent on target recognition by pairs of antibodies using in situ proximity ligation assays (PLA). Here we applied these DNA-assisted RCA techniques in capillary isoelectric focusing to resolve endogenous signaling transducers and isoforms along vascular endothelial growth factor (VEGF) signaling pathways at concentrations too low to be detected in standard assays. We also demonstrate background rejection and enhanced specificity when protein detection depended on binding by pairs of antibodies using in situ PLA, compared to assays where each antibody preparation was used on its own.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Anti-CDC34 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-S100A6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-AKR1B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)