Skip to Content
Merck
All Photos(2)

Key Documents

DUO92006

Sigma-Aldrich

Duolink® In Situ PLA® Probe Anti-Goat MINUS

Affinity purified Donkey anti-Goat IgG (H+L)

Synonym(s):

in situ Proximity Ligation Assay Kit, Protein Protein Interaction Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

donkey (polyclonal)

Quality Level

antibody form

affinity purified immunoglobulin (secondary antibody)

antibody product type

primary antibodies

product line

Duolink®

species reactivity

goat

technique(s)

immunofluorescence: suitable
proximity ligation assay: suitable

suitability

suitable for brightfield
suitable for fluorescence

shipped in

wet ice

storage temp.

2-8°C

Application

Duolink®proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.

This product can be applied to both the Duolink® In Situ Fluorescence Protocol and the Duolink® In Situ Brightfield Protocol depending on the detection reagents used.

Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.

To perform a complete Duolink® PLA in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. Other necessary reagents include a pair of PLA probes from different species (one PLUS and one MINUS), detection reagents, wash buffers, and mounting medium. Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.HRP is also available for brightfield detection.
Specificity
PLA probe anti-Goat reacts with whole molecule goat IgG and the light chains of other goat immunoglobulin?s. The PLA probe anti-Goat may cross-react with sheep antibodies, but has minimal cross reactivity with chicken, guinea pig, Syrian hamster, horse, human, mouse,rabbit, and rat serum proteins. A PLUS probe of a different species must be used simultaneously with this product. See our Product Selection Guide for more information.

Application Note
Two primary antibodies raised in different species are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC) or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink®PLA in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list

Features and Benefits

  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Single molecule sensitivity due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Quicker and simpler than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Components

This product is comprised of the following:
  • 5x PLA Probe Anti-Goat MINUS - Donkey anti-goat secondary antibody conjugated to oligonucleotide MINUS
  • 1x Blocking Solution - Reagent for blocking of the sample
  • 1x Antibody Diluent - For dilution of PLA probes and primary antibodies
See datasheet for more information.

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofan Li et al.
PLoS pathogens, 13(3), e1006249-e1006249 (2017-03-02)
Trials to reintroduce chloroquine into regions of Africa where P. falciparum has regained susceptibility to chloroquine are underway. However, there are long-standing concerns about whether chloroquine increases lytic-replication of Epstein-Barr virus (EBV), thereby contributing to the development of endemic Burkitt
Xiaofan Li et al.
Journal of virology, 93(17) (2019-06-14)
Herpesviruses are ubiquitous, and infection by some, like Epstein-Barr virus (EBV), is nearly universal. To persist, EBV must periodically switch from a latent to a replicative/lytic phase. This productive phase is responsible for most herpesvirus-associated diseases. EBV encodes a latency-to-lytic
Roberto Di Maio et al.
Science translational medicine, 10(451) (2018-07-27)
Missense mutations in leucine-rich repeat kinase 2 (LRRK2) cause familial Parkinson's disease (PD). However, a potential role of wild-type LRRK2 in idiopathic PD (iPD) remains unclear. Here, we developed proximity ligation assays to assess Ser1292 phosphorylation of LRRK2 and, separately
M Aubele et al.
British journal of cancer, 103(5), 663-667 (2010-08-12)
Protein tyrosine kinase 6 (PTK6; breast tumour kinase) is overexpressed in up to 86% of the invasive breast cancers, and its association with the oncoprotein human epidermal growth factor receptor 2 (HER2) was shown in vitro by co-precipitation. Furthermore, expression
Kan V Lu et al.
Cancer cell, 22(1), 21-35 (2012-07-14)
Inhibition of VEGF signaling leads to a proinvasive phenotype in mouse models of glioblastoma multiforme (GBM) and in a subset of GBM patients treated with bevacizumab. Here, we demonstrate that vascular endothelial growth factor (VEGF) directly and negatively regulates tumor

Articles

Support information including tips and tricks, frequently asked questions, and basic troubleshooting.

Things to consider for preparation, setup and execution of the Duolink® assay protocol

Protocols

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Duolink® PLA reagents enable brightfield detection and quantification of proteins and interactions in tissue samples.

Related Content

Applications to detect, quantify and visualize protein-protein interactions, post-translational modifications and low expression protein detection using proximity ligation assay

Questions

1–4 of 4 Questions  
  1. What epitope on the antibody is recognized by the Duolink PLA Probes?

    1 answer
    1. The Duolink PLA Probes offered are donkey polyclonal antibodies directed to the whole molecule (heavy and light chains).  They are affinity purified.  The same polyclonal antibody is conjugated to the Plus and Minus Oligos. The Plus and Minus probes can be used together (to detect a single antibody) since they can recognize multiple sites.  The anti-mouse IgG PLA labeled antibodies may cross-react with rat but this is not consistent. Do not use this product with rat antibodies.  It is best to label anti-rat secondary antibodies with the Duolink Probemaker kit.The anti-goat IgG PLA labeled antibodies have cross-reactivity with sheep IgG and can be recommended for this application.

      Helpful?

  2. Has Duolink been used for detection of changes in protein expression?

    1 answer
    1. The following reference cites the use of Duolink to detect changes in protein expression in Human. Raizer et al., Cancer 2010;116:5297-305

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How many reactions can be performed with the Duolink® In Situ Starter Kit?

    1 answer
    1. All of the starter kits for the Duolink product line will perform 30 reactions using 40 ul per reaction.    DUO92101 - Duolink® In Situ Red Starter Kit Mouse/Rabbit    DUO92102 - Duolink® In Situ Orange Starter Kit Mouse/Rabbit    DUO92103 - Duolink® In Situ Red Starter Kit Mouse/Goat    DUO92104 - Duolink® In Situ Orange Starter Kit Mouse/Goat    DUO92105 - Duolink® In Situ Red Starter Kit Goat/Rabbit    DUO92106 - Duolink® In Situ Orange Starter Kit Goat/Rabbit

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service