Sigma-Aldrich
Sodium thiosulfate solution, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiosulfate solution, 0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiosulfate solution, 0.025 M
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiosulfate solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Bisphenol A, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile
Pricing and availability is not currently available.