Merck
  • LIM-only protein FHL2 critically determines survival and radioresistance of pancreatic cancer cells.
LIM-only protein FHL2 critically determines survival and radioresistance of pancreatic cancer cells.
Cancer letters (2015-04-29)
Elisa Zienert, Iris Eke, Daniela Aust, Nils Cordes
ABSTRACT
Numerous factors determine the current poor prognosis of pancreatic ductal adenocarcinoma (PDAC). One of the greatest challenges to overcome is treatment resistance. Among a large repertoire of intrinsic resistance mechanisms, integrin-mediated cell adhesion to extracellular matrix (ECM) has been identified to be fundamental. Coalesced in focal adhesion complexes, integrins, receptor tyrosine kinases, protein kinases and adapter proteins mediate prosurvival signaling. Four and a half LIM domains protein 2 (FHL2) is one of these adapter proteins, which operates through protein-protein interactions and shows tumor-specific expression. Based on this, we investigated FHL2 expression in PDAC specimens and three-dimensionally grown cell lines and how FHL2 mechanistically contributes to cell survival, cell cycling and radiation resistance. PDAC exhibited a significantly increased and heterogeneous FHL2 expression. Upon FHL2 depletion, pancreatic cancer cell lines showed significantly decreased cell survival, proliferation and radioresistance as well as enhanced apoptosis and MEK/ERK signaling and cyclin D1, E, A and B1 expression were strongly induced. Targeting of FHL2 and MEK1 was similarly effective than FHL2 depletion alone, suggesting MEK1 as a downstream signaling mediator of FHL2. Taken together, our results provide evidence for the importance of the focal adhesion protein FHL2 in pancreatic cancer cell survival, proliferation and radiosensitivity.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Map2k1
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP2K1
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
SAFC
Sodium deoxycholate