Merck
  • New ester based gemini surfactants: the effect of different cationic headgroups on micellization properties and viscosity of aqueous micellar solution.
New ester based gemini surfactants: the effect of different cationic headgroups on micellization properties and viscosity of aqueous micellar solution.
Physical chemistry chemical physics : PCCP (2015-07-07)
Avinash Bhadani, Misako Tani, Takeshi Endo, Kenichi Sakai, Masahiko Abe, Hideki Sakai
ABSTRACT
A new series of ester functionalized cationic gemini surfactants having different cationic headgroups (i.e. piperidinium, pyrrolidinium, morpholinium and quaternary ammonium) have been synthesized and characterized using NMR and Mass spectroscopy. These new gemini surfactants were investigated for their micellization and viscosity properties using surface tension, conductivity, fluorescence and rheology thechniques. The physicochemical properties of the aqueous surfactant system were influenced by polarity, size and the nature of cationic headgroups as the surface, thermodynamic and viscosity properties of these gemini surfactants were found to be dependent on the type of cationic headgroup. The current research finding establishes the structure-property relationship of the surfactant molecule specifically taking into account the dominant role displayed by the nature of the cationic headgroup.
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Piperidine, SAJ first grade, ≥99.0%
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Bromoacetic acid, reagent grade, 97%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Diethylamine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Diethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Morpholine, polymer-bound, 200-400 mesh, extent of labeling: 2.75-3.25 mmol/g loading, 1 % cross-linked
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Diethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
StratoSpheres PL-MPH resin, 50-100 mesh, extent of labeling: 3.0-4.0 mmol/g loading, 1 % cross-linked
Sigma-Aldrich
1-Bromododecane, 97%