Merck
  • Reactivated herpes simplex infection increases the risk of Alzheimer's disease.
Reactivated herpes simplex infection increases the risk of Alzheimer's disease.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2014-07-22)
Hugo Lövheim, Jonathan Gilthorpe, Rolf Adolfsson, Lars-Göran Nilsson, Fredrik Elgh
ABSTRACT
Previous studies have suggested a link between herpes simplex virus (HSV) type 1 and the development of Alzheimer's disease (AD). The present analysis included 3432 persons (53.9% women, mean age at inclusion 62.7 ± 14.4 years) with a mean follow-up time of 11.3 years. The number of incident AD cases was 245. Serum samples were analyzed for anti-HSV antibodies (immunoglobulin (Ig)G and IgM) by enzyme-linked immunosorbent assays. The presence of anti-HSV IgG antibodies was not associated with an increased risk for AD, controlled for age and sex (hazard ratio, HR, 0.993, P = .979). However, the presence of anti-HSV IgM at baseline was associated with an increased risk of developing AD (HR 1.959, P = .012). Positivity for anti-HSV IgM, a sign of reactivated infection, was found to almost double the risk for AD, whereas the presence of anti-HSV IgG antibodies did not affect the risk.
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)