Merck
  • Structural insights into the assembly of the histone deacetylase-associated Sin3L/Rpd3L corepressor complex.
Structural insights into the assembly of the histone deacetylase-associated Sin3L/Rpd3L corepressor complex.
Proceedings of the National Academy of Sciences of the United States of America (2015-07-01)
Michael D Clark, Ryan Marcum, Richard Graveline, Clarence W Chan, Tao Xie, Zhonglei Chen, Yujia Ding, Yongbo Zhang, Alfonso Mondragón, Gregory David, Ishwar Radhakrishnan
ABSTRACT
Acetylation is correlated with chromatin decondensation and transcriptional activation, but its regulation by histone deacetylase (HDAC)-bearing corepressor complexes is poorly understood. Here, we describe the mechanism of assembly of the mammalian Sin3L/Rpd3L complex facilitated by Sds3, a conserved subunit deemed critical for proper assembly. Sds3 engages a globular, helical region of the HDAC interaction domain (HID) of the scaffolding protein Sin3A through a bipartite motif comprising a helix and an adjacent extended segment. Sds3 dimerizes through not only one of the predicted coiled-coil motifs but also, the segment preceding it, forming an ∼ 150-Å-long antiparallel dimer. Contrary to previous findings in yeast, Sin3A rather than Sds3 functions in recruiting HDAC1 into the complex by engaging the latter through a highly conserved segment adjacent to the helical HID subdomain. In the resulting model for the ternary complex, the two copies of the HDACs are situated distally and dynamically because of a natively unstructured linker connecting the dimerization domain and the Sin3A interaction domain of Sds3; these features contrast with the static organization described previously for the NuRD (nucleosome remodeling and deacetylase) complex. The Sds3 linker features several conserved basic residues that could potentially maintain the complex on chromatin by nonspecific interactions with DNA after initial recruitment by sequence-specific DNA-binding repressors.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Monoclonal Anti-c-Myc antibody produced in mouse, clone 9E10, ascites fluid
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC