Merck
  • Postsynthetic conjugation of RNA to carboxylate and dicarboxylate molecules.
Postsynthetic conjugation of RNA to carboxylate and dicarboxylate molecules.
Nucleosides, nucleotides & nucleic acids (2015-10-01)
Yossi Shemesh, Eylon Yavin
ABSTRACT
Carboxylates and dicarboxylates are important phosphate mimics. Herein, we present a simple synthetic route for the preparation of RNA carboxylate/dicarboxylate conjugates, starting from suitably protected NH2- and COOH-containing molecules that are coupled to the RNA on the solid support. The key point in our method was the use of trimethylsilylethanol (TMSE-OH) protecting group, which is removed simultaneously with the silyl protecting group on the 2'-OH of the RNA ribose (e.g. t-Butyldimethylsilyl) during the final RNA cleavage/deprotection steps. The usefulness of this method was demonstrated by preparing different RNA-phosphate mimics oligos.
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
CDI, reagent grade
Sigma-Aldrich
Palladium hydroxide on carbon, extent of labeling: 20 wt. % loading (dry basis), matrix carbon, wet support
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Palladium hydroxide on carbon, extent of labeling: 20 wt. % loading (dry basis), matrix carbon, wet support
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Hydrochloric acid solution, 1 M