Merck
Antioxidants in heat-processed koji and the production mechanisms.
Food chemistry (2015-05-16)
Kayu Okutsu, Yumiko Yoshizaki, Natsumi Ikeda, Tatsuro Kusano, Fumio Hashimoto, Kazunori Takamine
ABSTRACT
We previously developed antioxidative heat-processed (HP)-koji via two-step heating (55 °C/2days → 75 °C/3 days) of white-koji. In this study, we isolated antioxidants in HP-koji and investigated their formation mechanisms. The antioxidants were identified to be 5-hydroxymethyl furfural (HMF) and 5-(α-D-glucopyranosyloxymethyl)-2-furfural (GMF) based on nuclear magnetic resonance spectral analysis. HMF and GMF were not present in intact koji, but were formed by heating at 75 °C. As production of these antioxidants was more effective by two-step heating than by constant heating at 55 °C or 75 °C, we presumed that the antioxidant precursors are derived enzymatically at 55°C and that the antioxidants are formed subsequently by thermal reaction at 75 °C. The heating assay of saccharide solutions revealed glucose and isomaltose as HMF and GMF precursors, respectively, and thus the novel finding of GMF formation from isomaltose. Finally, HMF and GMF were effectively formed by two-step heating from glucose and isomaltose present in koji.
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, SAJ special grade