Merck
  • Drug-eluting microarrays to identify effective chemotherapeutic combinations targeting patient-derived cancer stem cells.
Drug-eluting microarrays to identify effective chemotherapeutic combinations targeting patient-derived cancer stem cells.
Proceedings of the National Academy of Sciences of the United States of America (2015-07-01)
Matthew R Carstens, Robert C Fisher, Abhinav P Acharya, Elizabeth A Butterworth, Edward Scott, Emina H Huang, Benjamin G Keselowsky
ABSTRACT
A new paradigm in oncology establishes a spectrum of tumorigenic potential across the heterogeneous phenotypes within a tumor. The cancer stem cell hypothesis postulates that a minute fraction of cells within a tumor, termed cancer stem cells (CSCs), have a tumor-initiating capacity that propels tumor growth. An application of this discovery is to target this critical cell population using chemotherapy; however, the process of isolating these cells is arduous, and the rarity of CSCs makes it difficult to test potential drug candidates in a robust fashion, particularly for individual patients. To address the challenge of screening drug libraries on patient-derived populations of rare cells, such as CSCs, we have developed a drug-eluting microarray, a miniaturized platform onto which a minimal quantity of cells can adhere and be exposed to unique treatment conditions. Hundreds of drug-loaded polymer islands acting as drug depots colocalized with adherent cells are surrounded by a nonfouling background, creating isolated culture environments on a solid substrate. Significant results can be obtained by testing <6% of the cells required for a typical 96-well plate. Reliability was demonstrated by an average coefficient of variation of 14% between all of the microarrays and 13% between identical conditions within a single microarray. Using the drug-eluting array, colorectal CSCs isolated from two patients exhibited unique responses to drug combinations when cultured on the drug-eluting microarray, highlighting the potential as a prognostic tool to identify personalized chemotherapeutic regimens targeting CSCs.
Sigma-Aldrich
Cyclohexanol, SAJ first grade, ≥95.0%
Sigma-Aldrich
Cyclohexanol, JIS special grade, ≥98.0%
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Sigma-Aldrich
Cyclohexanol, ReagentPlus®, 99%
Sigma-Aldrich
Vinyl acetate, CP