Merck
  • Antioncogenic and Oncogenic Properties of Nrf2 in Arsenic-induced Carcinogenesis.
Antioncogenic and Oncogenic Properties of Nrf2 in Arsenic-induced Carcinogenesis.
The Journal of biological chemistry (2015-09-20)
Young-Ok Son, Poyil Pratheeshkumar, Ram Vinod Roy, John Andrew Hitron, Lei Wang, Sasidharan Padmaja Divya, Mei Xu, Jia Luo, Gang Chen, Zhuo Zhang, Xianglin Shi
ABSTRACT
Arsenic (As(3+)) is a carcinogen with considerable environmental and occupational relevancy. The present study shows that As(3+)-transformed human lung bronchial epithelial BEAS-2B cells (AsT cells) exhibit the property of apoptosis resistance. The level of basal reactive oxygen species (ROS) is very low in AsT cells in correlation with elevated expressions of both antioxidant enzymes and antiapoptotic proteins. Nuclear factor erythroid 2-related factor (Nrf2) and p62 are constitutively expressed. These two proteins up-regulate antioxidant enzymes and antiapoptotic proteins. The knockdown of Nrf2 or p62 by small interfering RNA (siRNA) enhanced both ROS levels and As(3+)-induced apoptosis in transformed cells. AsT cells have autophagy deficiency as evidenced by reduced formation of microtubule-associated protein 1 light chain 3 (LC3)-II, GFP-LC3 puncta, and autophagy flux. Results obtained using a soft agar assay and shRNA Nrf2-transfected cells show that Nrf2 plays an antioncogenic role before transformation, whereas this transcription factor plays an oncogenic role after transformation. In addition, depletion of Nrf2 by shRNA dramatically inhibited growth and proliferation of transformed cells. Furthermore, the Nrf2 protein levels and antiapoptotic and antioxidant enzyme levels are higher in lung adenocarcinoma than in normal tissues. Collectively, this study demonstrates that a constitutively high level of Nrf2 in AsT cells up-regulates the antioxidant proteins catalase and superoxide dismutase as well as the antiapoptotic proteins Bcl-2 and Bcl-xL. The final consequences are decreased ROS generation and increased apoptotic resistance, cell survival and proliferation, and tumorigenesis.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nfe2l2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gtf2h1
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dctn4
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
MISSION® esiRNA, targeting human GTF2H1
Sigma-Aldrich
MISSION® esiRNA, targeting human DCTN4