Merck
  • Influence of coffee genotype on bioactive compounds and the in vitro capacity to scavenge reactive oxygen and nitrogen species.
Influence of coffee genotype on bioactive compounds and the in vitro capacity to scavenge reactive oxygen and nitrogen species.
Journal of agricultural and food chemistry (2015-04-25)
Naira Poerner Rodrigues, Terezinha de Jesus Garcia Salva, Neura Bragagnolo
ABSTRACT
The influence of green coffee genotype on the bioactive compounds and the in vitro antioxidant capacity against the principal reactive oxygen (ROO(•), H2O2, HO(•), and HOCl) and nitrogen (NO(•) and ONOO(-)) species of biological relevance was investigated. This is the first report on the capacity of green coffee to scavenge H2O2, HOCl, and NO(•). Variations in the contents of total chlorogenic acids (22.9-37.9 g/100 g), cinnamoyl-amino acid conjugates (0.03-1.12 g/100 g), trigonelline (3.1-6.7 g/100 g), and caffeine (3.9-11.8 g/100 g) were found. Hydrophilic extracts of Coffea canephora and Coffea kapakata were the most potent scavengers of ROO(•), H2O2, HO(•), NO(•), and ONOO(-) due to their chlorogenic acid contents, which were, on average, 30% higher than those found in Coffea arabica and Coffea racemosa. The results showed that genotype is a determinant characteristic in the bioactive compound contents and consequently in the antioxidant capacity of green coffee.
Sigma-Aldrich
Sodium hypochlorite solution, CP
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Sodium hypochlorite solution, CP
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%