Merck
  • Semaglutide, a once-weekly human GLP-1 analog, does not reduce the bioavailability of the combined oral contraceptive, ethinylestradiol/levonorgestrel.
Semaglutide, a once-weekly human GLP-1 analog, does not reduce the bioavailability of the combined oral contraceptive, ethinylestradiol/levonorgestrel.
Journal of clinical pharmacology (2014-12-06)
Christoph Kapitza, Leszek Nosek, Lene Jensen, Helle Hartvig, Christine B Jensen, Anne Flint
ABSTRACT
The effect of semaglutide, a once-weekly human glucagon-like peptide-1 (GLP-1) analog in development for type 2 diabetes (T2D), on the bioavailability of a combined oral contraceptive was investigated. Postmenopausal women with T2D (n = 43) on diet/exercise ± metformin received ethinylestradiol (0.03 mg)/levonorgestrel (0.15 mg) once daily for 8 days before (semaglutide-free) and during (steady-state 1.0 mg) semaglutide treatment (subcutaneous once weekly; dose escalation: 0.25 mg 4 weeks; 0.5 mg 4 weeks; 1.0 mg 5 weeks). Bioequivalence of oral contraceptives was established if 90%CI for the ratio of pharmacokinetic parameters during semaglutide steady-state and semaglutide-free periods was within prespecified limits (0.80-1.25). The bioequivalence criterion was met for ethinylestradiol area under the curve (AUC0-24 h ) for semaglutide steady-state/semaglutide-free; 1.11 (1.06-1.15). AUC0-24 h was 20% higher for levonorgestrel at semaglutide steady-state vs. semaglutide-free (1.20 [1.15-1.26]). Cmax was within bioequivalence criterion for both contraceptives. Reductions (mean ± SD) in HbA1c (-1.1 ± 0.6%) and weight (-4.3 ± 3.1 kg) were observed. Semaglutide pharmacokinetics were compatible with once-weekly dosing; the semaglutide dose and dose-escalation regimen were well tolerated. Adverse events, mainly gastrointestinal, were mild to moderate in severity. Asymptomatic increases in mean amylase and lipase were observed. Three subjects had elevated alanine aminotransferase levels ≥3x the upper limit of normal during semaglutide/oral contraceptive coadministration, which were reported as adverse events, but resolved during follow-up. Semaglutide did not reduce the bioavailability of ethinylestradiol and levonorgestrel.
Product Description
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
D(−)-Norgestrel, analytical standard
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, British Pharmacopoeia (BP) Assay Standard
Levonorgestrel for system suitability 2, European Pharmacopoeia (EP) Reference Standard
Levonorgestrel, European Pharmacopoeia (EP) Reference Standard
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Levonorgestrel, United States Pharmacopeia (USP) Reference Standard