Merck
  • Sorafenib resistance and JNK signaling in carcinoma during extracellular matrix stiffening.
Sorafenib resistance and JNK signaling in carcinoma during extracellular matrix stiffening.
Biomaterials (2014-04-15)
Thuy V Nguyen, Marianne Sleiman, Timothy Moriarty, William G Herrick, Shelly R Peyton
ABSTRACT
Tumor progression is coincident with mechanochemical changes in the extracellular matrix (ECM). We hypothesized that tumor stroma stiffening, alongside a shift in the ECM composition from a basement membrane-like microenvironment toward a dense network of collagen-rich fibers during tumorigenesis, confers resistance to otherwise powerful chemotherapeutics. To test this hypothesis, we created a high-throughput drug screening platform based on our poly(ethylene glycol)-phosphorylcholine (PEG-PC) hydrogel system, and customized it to capture the stiffness and integrin-binding profile of in vivo tumors. We report that the efficacy of a Raf kinase inhibitor, sorafenib, is reduced on stiff, collagen-rich microenvironments, independent of ROCK activity. Instead, sustained activation of JNK mediated this resistance, and combining a JNK inhibitor with sorafenib eliminated stiffness-mediated resistance in triple negative breast cancer cells. Surprisingly, neither ERK nor p38 appears to mediate sorafenib resistance, and instead, either ERK or p38 inhibition rescued sorafenib resistance during JNK inhibition, suggesting negative crosstalk between these signaling pathways on stiff, collagen-rich environments. Overall, we discovered that β1 integrin and its downstream effector JNK mediate sorafenib resistance during tumor stiffening. These results also highlight the need for more advanced cell culture platforms, such as our high-throughput PEG-PC system, with which to screen chemotherapeutics.
Product Description
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Propyl methacrylate, contains ~200 ppm MEHQ, 97%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture