Merck
  • Subnormothermic ex vivo liver perfusion reduces endothelial cell and bile duct injury after donation after cardiac death pig liver transplantation.
Subnormothermic ex vivo liver perfusion reduces endothelial cell and bile duct injury after donation after cardiac death pig liver transplantation.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society (2014-09-03)
Jan M Knaak, Vinzent N Spetzler, Nicolas Goldaracena, Markus U Boehnert, Fateh Bazerbachi, Kristine S Louis, Oyedele A Adeyi, Leonid Minkovich, Paul M Yip, Shaf Keshavjee, Gary A Levy, David R Grant, Nazia Selzner, Markus Selzner
ABSTRACT
An ischemic-type biliary stricture (ITBS) is a common feature after liver transplantation using donation after cardiac death (DCD) grafts. We compared sequential subnormothermic ex vivo liver perfusion (SNEVLP; 33°C) with cold storage (CS) for the prevention of ITBS in DCD liver grafts in pig liver transplantation (n = 5 for each group). Liver grafts were stored for 10 hours at 4°C (CS) or preserved with combined 7-hour CS and 3-hour SNEVLP. Parameters of hepatocyte [aspartate aminotransferase (AST), international normalized ratio (INR), factor V, and caspase 3 immunohistochemistry], endothelial cell (EC; CD31 immunohistochemistry and hyaluronic acid), and biliary injury and function [alkaline phosphatase (ALP), total bilirubin, and bile lactate dehydrogenase (LDH)] were determined. Long-term survival (7 days) after transplantation was similar between the SNEVLP and CS groups (60% versus 40%, P = 0.13). No difference was observed between SNEVLP- and CS-treated animals with respect to the peak of serum INR, factor V, or AST levels within 24 hours. CD31 staining 8 hours after transplantation demonstrated intact EC lining in SNEVLP-treated livers (7.3 × 10(-4) ± 2.6 × 10(-4) cells/μm(2)) but not in CS-treated livers (3.7 × 10(-4) ± 1.3 × 10(-4) cells/μm(2) , P = 0.03). Posttransplant SNEVLP animals had decreased serum ALP and serum bilirubin levels in comparison with CS animals. In addition, LDH in bile fluid was lower in SNEVLP pigs versus CS pigs (14 ± 10 versus 60 ± 18 μmol/L, P = 0.02). Bile duct histology revealed severe bile duct necrosis in 3 of 5 animals in the CS group but none in the SNEVLP group (P = 0.03). Sequential SNEVLP preservation of DCD grafts reduces bile duct and EC injury after liver transplantation.
Sigma-Aldrich
N-Acetyl-L-cysteine, SAJ special grade, 98.0-102.0%
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
BQ-123, ≥99%, sodium salt, lyophilized powder
Sevoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Supelco
Metronidazole, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, analytical standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al