Merck
  • Circuit formation and function in the olfactory bulb of mice with reduced spontaneous afferent activity.
Circuit formation and function in the olfactory bulb of mice with reduced spontaneous afferent activity.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-01-09)
Paolo Lorenzon, Nelly Redolfi, Michael J Podolsky, Ilaria Zamparo, Sira Angela Franchi, Gianluca Pietra, Anna Boccaccio, Anna Menini, Venkatesh N Murthy, Claudia Lodovichi
ABSTRACT
The type of neuronal activity required for circuit development is a matter of significant debate. We addressed this issue by analyzing the topographic organization of the olfactory bulb in transgenic mice engineered to have very little afferent spontaneous activity due to the overexpression of the inwardly rectifying potassium channel Kir2.1 in the olfactory sensory neurons (Kir2.1 mice). In these conditions, the topography of the olfactory bulb was unrefined. Odor-evoked responses were readily recorded in glomeruli with reduced spontaneous afferent activity, although the functional maps were coarser than in controls and contributed to altered olfactory discrimination behavior. In addition, overexpression of Kir2.1 in adults induced a regression of the already refined connectivity to an immature (i.e., coarser) status. Our data suggest that spontaneous activity plays a critical role not only in the development but also in the maintenance of the topography of the olfactory bulb and in sensory information processing.
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzaldehyde, SAJ special grade, ≥98.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
(±)-Sulfinpyrazone, analytical standard
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Geraniol, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%