Merck
  • CYP2C19*17 increases clopidogrel-mediated platelet inhibition but does not alter the pharmacokinetics of the active metabolite of clopidogrel.
CYP2C19*17 increases clopidogrel-mediated platelet inhibition but does not alter the pharmacokinetics of the active metabolite of clopidogrel.
Clinical and experimental pharmacology & physiology (2014-08-15)
Rasmus Steen Pedersen, Flemming Nielsen, Tore Bjerregaard Stage, Pernille Just Vinholt, Alaa Bilal el Achwah, Per Damkier, Kim Brosen
ABSTRACT
The aim of the present study was to determine the impact of CYP2C19*17 on the pharmacokinetics and pharmacodynamics of the active metabolite of clopidogrel and the pharmacokinetics of proguanil. Thus, we conducted an open-label two-phase cross-over study in 31 healthy male volunteers (11 CYP2C19*1/*1, 11 CYP2C19*1/*17 and nine CYP2C19*17/*17). In Phase A, the pharmacokinetics of the derivatized active metabolite of clopidogrel (CAMD) and platelet function were determined after administration of a single oral dose of 600 mg clopidogrel (Plavix; Sanofi-Avensis, Horsholm, Denmark). In Phase B, the pharmacokinetics of proguanil and its metabolites cycloguanil and 4-chlorphenylbiguanide (4-CPB) were determined in 29 of 31 subjects after a single oral dose of 200 mg proguanil given as the combination drug Malarone (GlaxoSmithKline Pharma, Brondby, Denmark). Significant correlations were found between the area under the time-concentration curve (AUC0-∞ ) of CAMD and both the absolute ADP-induced P2Y12 receptor-activated platelet aggregation (r = -0.60, P = 0.0007) and the percentage inhibition of aggregation (r = 0.59, P = 0.0009). In addition, the CYP2C19*17/*17 and CYP2C19*1/*17 genotype groups had significantly higher percentage inhibition of platelet aggregation compared with the CYP2C19*1/*1 subjects (geometric mean percentage inhibition of 84%, 73% and 63%, respectively; P = 0.014). Neither the absolute ADP-induced P2Y12 receptor-activated platelet aggregation, exposure to CAMD nor the pharmacokinetic parameters of proguanil, cycloguanil and 4-CPB exhibited any significant differences among the genotype groups. In conclusion, carriers of CYP2C19*17 exhibit higher percentage inhibition of platelet aggregation, but do not have significantly lower absolute P2Y12 receptor-activated platelet aggregation or higher exposure to the active metabolite after a single oral administration of 600 mg clopidogrel.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Atovaquone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Proguanil hydrochloride, ≥95% (HPLC)
Sigma-Aldrich
Atovaquone, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Atovaquone, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%