Merck
  • A method to identify and validate mitochondrial modulators using mammalian cells and the worm C. elegans.
A method to identify and validate mitochondrial modulators using mammalian cells and the worm C. elegans.
Scientific reports (2014-06-14)
Pénélope A Andreux, Laurent Mouchiroud, Xu Wang, Virginija Jovaisaite, Adrienne Mottis, Sabrina Bichet, Norman Moullan, Riekelt H Houtkooper, Johan Auwerx
ABSTRACT
Mitochondria are semi-autonomous organelles regulated by a complex network of proteins that are vital for many cellular functions. Because mitochondrial modulators can impact many aspects of cellular homeostasis, their identification and validation has proven challenging. It requires the measurement of multiple parameters in parallel to understand the exact nature of the changes induced by such compounds. We developed a platform of assays scoring for mitochondrial function in two complementary models systems, mammalian cells and C. elegans. We first optimized cell culture conditions and established the mitochondrial signature of 1,200 FDA-approved drugs in liver cells. Using cell-based and C. elegans assays, we further defined the metabolic effects of two pharmacological classes that emerged from our hit list, i.e. imidazoles and statins. We found that these two drug classes affect respiration through different and cholesterol-independent mechanisms in both models. Our screening strategy enabled us to unequivocally identify compounds that have toxic or beneficial effects on mitochondrial activity. Furthermore, the cross-species approach provided novel mechanistic insight and allowed early validation of hits that act on mitochondrial function.
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Simvastatin, analytical standard
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
USP
Miconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Bifonazole for system suitability, European Pharmacopoeia (EP) Reference Standard
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Miconazole, European Pharmacopoeia (EP) Reference Standard
Tioconazole for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Tioconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tioconazole, 97%
Econazole, European Pharmacopoeia (EP) Reference Standard