Merck
DNAJB1 destabilizes PDCD5 to suppress p53-mediated apoptosis.
Cancer letters (2014-12-03)
Xiandan Cui, Hyo-Kyoung Choi, Young-Seok Choi, Soo-Yeon Park, Gi-Jun Sung, Yoo-Hyun Lee, Jeongmin Lee, Woo Jin Jun, Kyungsup Kim, Kyung-Chul Choi, Ho-Geun Yoon
ABSTRACT
Although PDCD5 promotes p53-mediated apoptosis in various cancers, little is known about PDCD5 regulation. We recently found that DNAJB1 interacts with PDCD5 and induces the ubiquitin-dependent proteasomal degradation of PDCD5, thereby inhibiting p53-mediated apoptosis. To investigate these novel roles for PDCD5 and DNAJB1, we performed DNAJB1 mapping with PDCD5. PDCD5 specifically binds to the DNAJB1-D5 domain (Δ180-210), which was found to be essential for the stabilization of PDCD5. Further study showed that DNAJB1 post-translationally regulates PDCD5 stability. DNAJB1 ubiquitinated PDCD5 via a ubiquitin-mediated pathway. In human lung A549 cancer cells, DNAJB1 promoted the ubiquitination and degradation of PDCD5 and inhibited p53 activation. However, DNAJB1 knockdown in A549 cells increased the etoposide-induced activation of the p53-mediated apoptosis pathway and repressed cancer cell growth. Because this function was p53 dependent, DNAJB1 depletion increased the expression of p53-targeted apoptosis genes. In conclusion, we screened a novel PDCD5-associating protein, DNAJB1, by yeast two-hybrid screening and provided evidences that DNAJB1 targets PDCD5 to suppress p53-dependent apoptosis of cancer cells. Thus, we identified DNAJB1 as a negative regulator of PDCD5-mediated apoptosis and found that the apoptosis network of PDCD5 regulates cancer cell death.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder