Merck
KPT-330 has antitumour activity against non-small cell lung cancer.
British journal of cancer (2014-06-20)
H Sun, N Hattori, W Chien, Q Sun, M Sudo, G L E-Ling, L Ding, S L Lim, S Shacham, M Kauffman, T Nakamaki, H P Koeffler
ABSTRACT
We investigated the biologic and pharmacologic activities of a chromosome region maintenance 1 (CRM1) inhibitor against human non-small cell lung cancer (NSCLC) cells both in vitro and in vivo. The in vitro and in vivo effects of a novel CRM1 inhibitor (KPT-330) for a large number of anticancer parameters were evaluated using a large panel of 11 NSCLC cell lines containing different key driver mutations. Mice bearing human NSCLC xenografts were treated with KPT-330, and tumour growth was assessed. KPT-330 inhibited proliferation and induced cell cycle arrest and apoptosis-related proteins in 11 NSCLC cells lines. Moreover, the combination of KPT-330 with cisplatin synergistically enhanced the cell kill of the NSCLC cells in vitro. Human NSCLC tumours growing in immunodeficient mice were markedly inhibited by KPT-330. Also, KPT-330 was effective even against NSCLC cells with a transforming mutation of either exon 20 of EGFR, TP53, phosphatase and tensin homologue, RAS or PIK3CA, suggesting the drug might be effective against a variety of lung cancers irrespective of their driver mutation. Our results support clinical testing of KPT-330 as a novel therapeutic strategy for NSCLC.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Pricing and availability is not currently available.
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Pricing and availability is not currently available.
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Propidium iodide solution
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Pricing and availability is not currently available.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Pricing and availability is not currently available.
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Rapamycin, VETRANAL®, analytical standard
Pricing and availability is not currently available.