Skip to Content
Merck
All Photos(1)

Documents

C2210000

Cisplatin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

cis-Diammineplatinum(II) dichloride, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cisplatin

manufacturer/tradename

EDQM

mp

270 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N.N.Cl[Pt]Cl

InChI

1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2

InChI key

LXZZYRPGZAFOLE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the Issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia. Their suitability for any other use is not guaranteed and is the sole responsibility of the user. This standard is not intended for human or animal use.
Established for the preparation of the below-given solutions as per European Pharmacopoeia:
  • Reference solutions in the identification using infrared absorption spectroscopy and thin-layer chromatography, testing of related substances, and assay using liquid chromatography (General text 2.2.29) of cisplatin, according to the monograph 0599
  • Reference solution (b) for the testing of related substances in carboplatin using liquid chromatography (General text 2.2.29), according to the monograph 1081

Biochem/physiol Actions

Potent platinum-based antineoplastic agent. Forms cytotoxic adducts with the DNA dinucleotide d(pGpG), inducing intrastrand cross-links.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carboplatin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 6104-6105 (2022)
Cisplatin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 2231-2232 (2009)
Michael F Milosevic et al.
International journal of cancer, 135(7), 1692-1699 (2013-08-02)
Radiotherapy (RT) with concurrent cisplatin (CRT) is standard treatment for locally advanced cervical cancer. However, not all patients benefit from the addition of cisplatin to RT alone. This study explored the value of pretreatment tumor interstitial fluid pressure (IFP) and
James Chih-Hsin Yang et al.
The Lancet. Oncology, 16(2), 141-151 (2015-01-16)
We aimed to assess the effect of afatinib on overall survival of patients with EGFR mutation-positive lung adenocarcinoma through an analysis of data from two open-label, randomised, phase 3 trials. Previously untreated patients with EGFR mutation-positive stage IIIB or IV
Yi-Long Wu et al.
The Lancet. Oncology, 14(8), 777-786 (2013-06-21)
The results of FASTACT, a randomised, placebo-controlled, phase 2 study, showed that intercalated chemotherapy and erlotinib significantly prolonged progression-free survival (PFS) in patients with advanced non-small-cell lung cancer. We undertook FASTACT-2, a phase 3 study in a similar patient population.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service