Merck
  • A functional and protein-protein interaction analysis of neuroepithelial cell transforming gene 1 in hepatocellular carcinoma.
A functional and protein-protein interaction analysis of neuroepithelial cell transforming gene 1 in hepatocellular carcinoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-08-13)
Ke Ye, Shi Chang, Jindong Li, Xinying Li, Yuedu Zhou, Zhiming Wang
ABSTRACT
In our previous study, we reported that the expression of neuroepithelial transforming gene 1 (NET1) was increased in hepatocellular carcinoma (HCC) tissues and suggested that NET1 may serve as a novel prognostic predictor and a useful target for HCC therapy. The aim of this study was to determine the role of NET1 in HCC by monitoring the proliferation, migration, invasion, and metastasis of HCC cells in which NET1 has been stably knocked down. Additionally, to provide targets for therapeutic intervention, we aimed to identify the protein which interacts with NET1. NET1 knockdown significantly reduced the migration, invasion, and metastasis of MHCC-97H cells by 44, 65, and 77%, respectively, while cell proliferation was not significantly altered following NET1 knockdown. During our efforts to find potential NET1 modulators through protein-protein interactions, we identified merlin as a NET1-binding protein in a yeast two-hybrid screen. The interaction between NET1 and merlin was confirmed by co-immunoprecipitation. We found that the levels of NET1 protein decreased along with the increase of merlin levels. Furthermore, we demonstrated that merlin facilitates NET1 ubiquitination via the ubiquitin-proteasome pathway. Taken together, this study suggests that NET1 plays an important role in HCC cell migration, invasion, and metastasis, which are key aspects of HCC progression. Furthermore, merlin may serve as a tumor suppressor for HCC by facilitating ubiquitination of NET1.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%