Merck
  • Polar organic chemical integrative samplers for pesticides monitoring: impacts of field exposure conditions.
Polar organic chemical integrative samplers for pesticides monitoring: impacts of field exposure conditions.
The Science of the total environment (2014-05-17)
Sophie Lissalde, Nicolas Mazzella, Patrick Mazellier
ABSTRACT
This study focuses on how Polar Organic Chemical Integrative Samplers (POCIS) work in real environmental conditions. A selection of 23 polar pesticides and 8 metabolites were investigated by exposure of triplicates of integrative samplers in two rivers in France for successive 14-day periods. The pesticides and metabolites were trapped not only in Oasis HLB sorbent but also in the polyethersulfone (PES) membrane of the POCIS. The distribution of pesticides depended on the molecular structure. The use of the Performance Reference Compound (PRC) is also discussed here. The impact of some environmental parameters and exposure setup on the transfer of pesticides in POCIS sorbent was studied: river flow rate, biofouling on membranes, sampler holding design and position in the stream. Results show a significant impact of river flow velocity on PRC desorption, especially for values higher than 4 cm·s(-1). Some fouling was observed on the PES membrane which could potentially have an impact on molecule accumulation in the POCIS. Finally, the positioning of the sampler in the river did not have significant effects on pesticide accumulation, when perpendicular exposures were used (sampler positioning in front of the water flow). The POCIS with PRC correction seems to be a suitable tool for estimating time-weighted average (TWA) concentrations, for all the molecules except for one of the nine pesticides analyzed in these two French rivers.
Product Description
Supelco
Chlorpyrifos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Diuron-d6, PESTANAL®, analytical standard
Supelco
Carbofuran-d3, PESTANAL®, analytical standard
Supelco
Chlorpyrifos, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Supelco
Atrazine-d5, PESTANAL®, analytical standard
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v), for copper determination
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Supelco
Carbaryl, PESTANAL®, analytical standard
Supelco
Simazine, PESTANAL®, analytical standard
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, for residue analysis
Supelco
Methomyl, PESTANAL®, analytical standard
Supelco
Metolachlor, PESTANAL®, analytical standard
Supelco
Simazine solution, 100 μg/mL in methanol, PESTANAL®, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%