Merck
  • Evaluation of an electrochemical biosensor for uric acid measurement in human whole blood samples.
Evaluation of an electrochemical biosensor for uric acid measurement in human whole blood samples.
Clinica chimica acta; international journal of clinical chemistry (2014-05-13)
Li-Ting Liao, Chi-Chih Liao, Chiu-Ching Liu, Ting-Ya Yang, Giueng-Chueng Wang
ABSTRACT
Uric acid measurement has become increasingly important, and electrochemically modified detection method based portable devices hold a dominant position in the market for point of care and self-monitoring of uric acid blood levels. However, there has been a lack of detailed performance evaluation of the electrochemical detection devices that are currently being used in professional health care facilities and for home self-monitoring of uric acid. A commercially available uric acid monitoring system that is chemically modified to reduce interference was evaluated via clinical evaluation for its performance and interference as compared to a centralized laboratory instrument. Precision was within ±3.1% for 3 levels of control solutions and whole blood samples. A range from 30 to 55% was acceptable for the measurement of hematocrit levels in whole blood samples. There was no interference for the potential substances at their high therapeutic levels. Hemolyzed samples of up to 75 g/l showed no interference with test results obtained by the BeneCheck system, while a -45.9% bias% was obtained during testing of the same samples by a spectrophotometer. Clinical evaluation showed that >95% of tests were within ±20% bias% compared to a centralized instrument in hospitals. The uric acid monitoring system was suitable for use in monitoring or screening uric acid concentration for home users or professionals.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, FCC, FG