Merck
  • Impact of the HIV integrase genetic context on the phenotypic expression and in vivo emergence of raltegravir resistance mutations.
Impact of the HIV integrase genetic context on the phenotypic expression and in vivo emergence of raltegravir resistance mutations.
The Journal of antimicrobial chemotherapy (2014-10-23)
Thi Thu Nga Nguyen, Sylvie Rato, Jean-Michel Molina, François Clavel, Constance Delaugerre, Fabrizio Mammano
ABSTRACT
HIV resistance to the integrase inhibitor raltegravir in treated patients is characterized by distinct resistance pathways. We hypothesize that differences in the in vivo dynamics of HIV resistance to raltegravir are due to the genetic context of the integrase present at baseline. We studied four patients whose viruses evolved towards different resistance pathways. The integrase baseline sequences were inserted into a reference clone. Primary resistance mutations were then introduced and their impact on viral replication capacity (RC) and resistance was measured. Patients A and B experienced emergence and persistence of mutation N155H under raltegravir therapy. In the integrase sequence from Patient A, N155H conferred potent resistance coupled with a lower impact on RC than Q148H. In Patient B, instead, selection of N155H could be explained by the dramatic loss of RC induced by the alternative Q148H mutation. In Patient C, N155H initially emerged and was later replaced by Q148H. In this integrase context, N155H resulted in higher RC but lower resistance than Q148H. In Patient D, Q148H rapidly emerged without appearance of N155H. This was the only patient for whom Q148H conferred higher RC and resistance than N155H. The emergence of different resistance mutations in patients was in full agreement with the impact of mutations in different baseline integrase contexts. Evolution towards different resistance genotypes is thus largely determined by the capacity of different integrase sequences present at baseline to minimize the effect of mutations on virus RC while allowing expression of resistance.
Sigma-Aldrich
2-Chlorophenol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Chlorophenol, 98%
Supelco
2-Chlorophenol, PESTANAL®, analytical standard
Sigma-Aldrich
2-Chlorophenol, ≥99%
Sigma-Aldrich
Chlorophenol Red-β-D-galactopyranoside, ≥90% (HPLC)