Skip to Content
Merck
All Photos(1)

Key Documents

05-3710

Sigma-Aldrich

2-Chlorophenol

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OH
CAS Number:
Molecular Weight:
128.56
Beilstein:
1905114
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

availability

available only in Japan

refractive index

n20/D 1.558 (lit.)

bp

175-176 °C (lit.)

mp

8 °C (lit.)

density

1.241 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1Cl

InChI

1S/C6H5ClO/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

ISPYQTSUDJAMAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengyang Wu et al.
Water research, 188, 116465-116465 (2020-10-14)
We created a hydrogen-based membrane palladium-film reactor (MPfR) by depositing palladium nanoparticles (PdNPs) on hollow-fiber membranes via autocatalytic hydrogenation to form a Pd-film. The MPfR was used for hydrodechlorination (HDC) of 2,4-dichlorophenol (2,4-DCP). HDC performances and mechanisms were systematically evaluated
Thi Thu Nga Nguyen et al.
The Journal of antimicrobial chemotherapy, 70(3), 731-738 (2014-10-23)
HIV resistance to the integrase inhibitor raltegravir in treated patients is characterized by distinct resistance pathways. We hypothesize that differences in the in vivo dynamics of HIV resistance to raltegravir are due to the genetic context of the integrase present
M A Barakat et al.
Environmental science and pollution research international, 20(6), 3751-3759 (2012-11-20)
Photocatalytic process has shown recently a great potential as an environmental friendly and clean remediation technology for organic pollutants in wastewater. This work described the synthesis of silver-based bimetallic nanoparticles using colloid chemistry and the subsequent immobilization onto titania to
Esther Martí et al.
The Science of the total environment, 409(14), 2707-2716 (2011-05-03)
Three chlorophenolic compounds (2-chlorophenol, 2,4,6-trichlorophenol, and pentachlorophenol) were tested to assess their effects on two soils with different properties: a granitic soil (Haplic Arenosol) and a calcareous one (Calcaric Regosol). Different concentrations of the pollutants (ranging from 0.001 to 10,000
Mohammad Zain Khan et al.
Journal of hazardous materials, 190(1-3), 222-228 (2011-04-07)
2-Chlorophenol (2-CP) degrading aerobic granules were cultivated in a sequencing batch reactor (SBR) in presence of glucose. The organic loading rate (OLR) was increased from 6.9 to 9.7 kg COD m(-3)d(-1) (1150-1617 mg L(-1)COD per cycle) during the experiment. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service