Merck
  • Feeding protein supplements in alfalfa hay-based lactation diets improves nutrient utilization, lactational performance, and feed efficiency of dairy cows.
Feeding protein supplements in alfalfa hay-based lactation diets improves nutrient utilization, lactational performance, and feed efficiency of dairy cows.
Journal of dairy science (2014-09-30)
K Neal, J-S Eun, A J Young, K Mjoun, J O Hall
ABSTRACT
Due to the increasing cost of soybean meal and concerns of excess N being excreted into the environment, new protein supplements have been developed. Two products that have shown potential in increasing N utilization efficiency are slow-release urea (SRU; Optigen; Alltech Inc., Nicholasville, KY) and ruminal-escape protein derived from yeast (YMP; DEMP; Alltech Inc.). The objective of this study was to assess the effects of feeding these 2 supplements in alfalfa hay-based [45.7% of forage dietary dry matter (DM)] dairy diets on nutrient utilization, feed efficiency, and lactational performance of dairy cows. Twelve multiparous dairy cows were used in a triple 4 × 4 Latin square design with one square consisting of ruminally cannulated cows. Treatments included (1) control, (2) SRU-supplemented total mixed ration (SRUT), (3) YMP-supplemented total mixed ration (YMPT), and (4) SRU- and YMP-supplemented total mixed ration (SYT). The control consisted only of a mixture of soybean meal and canola meal in a 50:50 ratio. The SRU and the YMP were supplemented at 0.49 and 1.15% DM, respectively. The experiment consisted of 4 periods lasting 28 d each (21 d of adaptation and 7 d of sampling). Cows fed YMPT and SYT had decreased intake of DM, and all supplemented treatments had lower crude protein intake compared with those fed the control. Milk yield tended to have the greatest increase in YMPT compared with the control (41.1 vs. 39.7 kg/d) as well as a tendency for increased milk fat and protein yields. Feed efficiencies based on yields of milk, 3.5% fat-corrected milk, and energy-corrected milk increased at 10 to 16% due to protein supplementation. Cows fed protein supplements partitioned less energy toward body weight gain, but tended to partition more energy toward milk production. Efficiency of use of feed N to milk N increased by feeding SRUT and YMPT, and milk N-to-manure N ratio increased with YMPT. Overall results from this experiment indicate that replacing the mixture of soybean meal and canola meal with SRU and YMP in alfalfa hay-based dairy diets can be a good approach to improve nutrient utilization efficiencies in lactating dairy cows.
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Diethyl methylphosphonate, 97%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urea, analytical standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium sulfite, BioUltra, anhydrous, ≥98% (RT)
Sigma-Aldrich
Allantoin, ≥98.0% (N)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium sulfite, BioXtra, ≥98%
Allantoin, European Pharmacopoeia (EP) Reference Standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Sodium sulfite, JIS special grade, ≥97.0%