Merck
  • A novel quantification method for analysis of twenty natural amino acids in human serum based on N-phosphorylation labeling using reversed-phase liquid chromatography-tandem mass spectrometry.
A novel quantification method for analysis of twenty natural amino acids in human serum based on N-phosphorylation labeling using reversed-phase liquid chromatography-tandem mass spectrometry.
Analytica chimica acta (2014-07-01)
Xiaowu Chen, Dan Gao, Feng Liu, Xiang Gao, Shujuan Wang, Yufen Zhao, Hongxia Liu, Yuyang Jiang
ABSTRACT
A novel method based on the strategy of N-phosphorylation labeling is described for quantification of twenty natural amino acids in human serum by reversed-phase liquid chromatography-electrospray tandem mass spectrometry (RP-LC/ESI-MS). The derivatization reaction was easily performed in one-pot reaction under mild conditions within 30min. The reaction mixture was then evaporated to dryness, redissolved, desalted by C18 SPE. The twenty N-phosphoryl amino acids were separated on an RP-C18 column within 20min by isocratic elution (0.1% formic acid-acetonitrile, v/v 7:3). At the same time, multiple reaction monitoring (MRM) MS enabled quantitation of twenty natural amino with the LOD of 0.0005-0.15μM and LOQ of 0.0020-0.5μM in human serum. The linear range was from 0.025 to 25μM (except Cys and Trp) with R>0.99. The recovery range was determined to be 85.5-117.4% with the relative standard deviation (RSD) in the range of 1.3-13.9%. All twenty amino acids were successfully detected in human serum samples with the concentration from 5.7 to 577.9μM, which indicates potential of the developed method for determination of amino acids in complex biological samples, hence for screening of amino acid metabolite related diseases.
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300