Merck
  • Comparison of three-factor and four-factor prothrombin complex concentrates regarding reversal of the anticoagulant effects of rivaroxaban in healthy volunteers.
Comparison of three-factor and four-factor prothrombin complex concentrates regarding reversal of the anticoagulant effects of rivaroxaban in healthy volunteers.
Journal of thrombosis and haemostasis : JTH (2014-05-09)
M Levi, K T Moore, C F Castillejos, D Kubitza, S D Berkowitz, S Z Goldhaber, M Raghoebar, M R Patel, J I Weitz, J H Levy
ABSTRACT
Four-factor prothrombin complex concentrates (PCCs), which contain factor II, FVII, FIX, and FX, have shown the potential to reverse the anticoagulant effect of rivaroxaban in healthy volunteers. The purpose of this study was to determine whether a three-factor PCC, which contains little FVII, has a similar effect. We performed an open-label, single-center, parallel-group study comparing the effect of a three-factor PCC (Profilnine SD) with that of a four-factor PCC (Beriplex P/N) on the pharmacodynamics of rivaroxaban in 35 healthy volunteers. After receiving 4 days of rivaroxaban 20 mg twice daily to obtain supratherapeutic steady-state concentrations, volunteers were randomized to receive a single 50 IU kg(-1) bolus dose of four-factor PCC, three-factor PCC or saline 4 h after the morning dose of rivaroxaban on day 5, and the effects of these interventions on prothrombin time and thrombin generation were determined. Within 30 min, four-factor PCC reduced mean prothrombin time by 2.5-3.5 s, whereas three-factor PCC produced only a 0.6-1.0-s reduction. In contrast, three-factor PCC reversed rivaroxaban-induced changes in thrombin generation more than four-factor PCC. This study demonstrates the potential of both three-factor and four-factor PCCs to at least partially reverse the anticoagulant effects of rivaroxaban in healthy adults. The discrepant effects of the PCC preparations may reflect differences in the procoagulant components present in each.
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, for ion-selective electrodes, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%