All Photos(1)
About This Item
Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Recommended Products
form
liquid
availability
available only in Japan
concentration
0.025 M
SMILES string
Cl[Ca]Cl
InChI
1S/Ca.2ClH/h;2*1H/q+2;;/p-2
InChI key
UXVMQQNJUSDDNG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
J Philippe et al.
International journal of obesity (2005), 39(2), 295-302 (2014-06-04)
A significant proportion of severe familial forms of obesity remain genetically elusive. Taking advantage of our unique cohort of multigenerational obese families, we aimed to assess the contribution of rare mutations in 29 common obesity-associated genes to familial obesity, and
Mohsen Akbari et al.
Advanced functional materials, 24(26), 4060-4067 (2014-11-21)
The fabrication of cell-laden structures with anisotropic mechanical properties while having a precise control over the distribution of different cell types within the constructs is important for many tissue engineering applications. Automated textile technologies for making fabrics allow simultaneous control
Michael A Goren et al.
Nature communications, 5, 5115-5115 (2014-10-09)
Opsin, the rhodopsin apoprotein, was recently shown to be an ATP-independent flippase (or scramblase) that equilibrates phospholipids across photoreceptor disc membranes in mammalian retina, a process required for disc homoeostasis. Here we show that scrambling is a constitutive activity of
S Zhen et al.
Gene therapy, 22(5), 404-412 (2015-02-06)
The current therapies to treat hepatitis B virus (HBV) infection are limited. Recently, clustered regularly interspaced short palindromic repeat (CRISPR) systems, originally identified in bacteria and archaea, have been found to consist of an RNA-based adaptive immune system that degrades
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service