Merck
  • Surface properties of artificial tear film lipid layers: effects of wax esters.
Surface properties of artificial tear film lipid layers: effects of wax esters.
Investigative ophthalmology & visual science (2014-05-31)
Pipsa Kulovesi, Antti H Rantamäki, Juha M Holopainen
ABSTRACT
The tear film lipid layer is believed to stabilize the tear film and to retard evaporation. Based on previous simple in vitro studies, the evidence for the latter property is scarce. In this study, we used complex lipid mixtures including various wax esters to study their physical properties and evaporation retarding effect. Twelve samples of artificial tear film lipid layer mixtures composed of (L-α)-phosphatidylcholine, cholesterol oleate, and triglycerides were mixed with wax esters. A Langmuir balance was used to analyze the compressibility and rheological properties of these mixtures. In addition, a custom-built system was used for the evaporation studies used at 35°C. Lipid films were imaged with Brewster angle microscopy. None of the studied lipid mixtures decreased the evaporation rate. All lipid mixtures had similar compression isotherms and viscoelastic properties regardless of the wax ester species or its concentration. The results suggest that the overall properties of these mixtures are independent of individual lipid species and that these films are very cooperative and showed minor variation depending on the wax ester species. Brewster angle microscopy images revealed that the lipid films assembled into multiple layers. Wax ester-containing lipid mixtures resembling the tear film lipid layer are organized in a layered fashion so that amphiphilic lipids are adjacent to the aqueous phase and the nonpolar lipids are layered on top of these. This organization does not retard evaporation and raises overall questions about the role of lipids in the tear film.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Trilaurin, European Pharmacopoeia (EP) Reference Standard
Tricaprin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
1-Docosanol, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Tricaprylin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cholesteryl oleate, ≥98% (HPLC; detection at 205 nm)
Trimyristin, European Pharmacopoeia (EP) Reference Standard