Merck
Bright light-emitting diodes based on organometal halide perovskite.
Nature nanotechnology (2014-08-05)
Zhi-Kuang Tan, Reza Saberi Moghaddam, May Ling Lai, Pablo Docampo, Ruben Higler, Felix Deschler, Michael Price, Aditya Sadhanala, Luis M Pazos, Dan Credgington, Fabian Hanusch, Thomas Bein, Henry J Snaith, Richard H Friend
ABSTRACT
Solid-state light-emitting devices based on direct-bandgap semiconductors have, over the past two decades, been utilized as energy-efficient sources of lighting. However, fabrication of these devices typically relies on expensive high-temperature and high-vacuum processes, rendering them uneconomical for use in large-area displays. Here, we report high-brightness light-emitting diodes based on solution-processed organometal halide perovskites. We demonstrate electroluminescence in the near-infrared, green and red by tuning the halide compositions in the perovskite. In our infrared device, a thin 15 nm layer of CH3NH3PbI(3-x)Cl(x) perovskite emitter is sandwiched between larger-bandgap titanium dioxide (TiO2) and poly(9,9'-dioctylfluorene) (F8) layers, effectively confining electrons and holes in the perovskite layer for radiative recombination. We report an infrared radiance of 13.2 W sr(-1) m(-2) at a current density of 363 mA cm(-2), with highest external and internal quantum efficiencies of 0.76% and 3.4%, respectively. In our green light-emitting device with an ITO/PEDOT:PSS/CH3NH3PbBr3/F8/Ca/Ag structure, we achieved a luminance of 364 cd m(-2) at a current density of 123 mA cm(-2), giving external and internal quantum efficiencies of 0.1% and 0.4%, respectively. We show, using photoluminescence studies, that radiative bimolecular recombination is dominant at higher excitation densities. Hence, the quantum efficiencies of the perovskite light-emitting diodes increase at higher current densities. This demonstration of effective perovskite electroluminescence offers scope for developing this unique class of materials into efficient and colour-tunable light emitters for low-cost display, lighting and optical communication applications.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylammonium bromide
Sigma-Aldrich
Formamidinium iodide
Sigma-Aldrich
Phenylammonium iodide
Sigma-Aldrich
Methylammonium iodide
Sigma-Aldrich
Methylammonium iodide, 98%
Sigma-Aldrich
Guanidinium iodide, ≥99%
Sigma-Aldrich
Phenethylammonium iodide
Sigma-Aldrich
Ethylammonium Iodide
Sigma-Aldrich
Dimethylammonium iodide
Sigma-Aldrich
n-Propylammonium iodide
Sigma-Aldrich
Benzylammonium iodide
Sigma-Aldrich
n-Butylammonium iodide
Sigma-Aldrich
Acetamidinium iodide