Skip to Content
Merck
All Photos(2)

Key Documents

805823

Sigma-Aldrich

Ethylammonium Iodide

greener alternative

Synonym(s):

Ethanamine hydriodide, Greatcell Solar®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H8IN
CAS Number:
Molecular Weight:
173.00
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

193 °C

greener alternative category

SMILES string

CCN.I

InChI

1S/C2H7N.HI/c1-2-3;/h2-3H2,1H3;1H

InChI key

XFYICZOIWSBQSK-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Ethylammonium iodide (EAI) is an organic ammonium salt that can be used as an additive in the fabrication of perovskite-based photovoltaic devices. It is also used in the synthesis of nanocrystalline sensitizers for the development of light-absorbing materials.
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structure, and photovoltaic property of a nanocrystalline 2H perovskite-type novel sensitizer (CH 3 CH 2 NH 3) PbI 3
Im J, et al.
Nanoscale Research Letters, 7(1), 353-353 (2012)
High-performance and high-durability perovskite photovoltaic devices prepared using ethylammonium iodide as an additive
Hsu H, et al.
Journal of Material Chemistry A, 3(17), 9271-9277 (2015)
Wei-Yun Liang et al.
Optics express, 28(17), 24919-24927 (2020-09-11)
The nonlinear optical properties of low-dimensional polycrystalline halide perovskite films consisting of ethylammonium (EA) and butylammonium (BA) cations are investigated using Z-scan technique. Across the band-edge, two-dimensional (BA)2PbI4 exhibits a transition from saturable absorption (SA) to reverse-SA and its nonlinear
Stephen J Page et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 316(1), R38-R49 (2018-10-26)
Nicotine is an addictive drug that has broad effects throughout the brain. One site of action is the nucleus of the solitary tract (NTS), where nicotine initiates a stress response and modulates cardiovascular and gastric function through nicotinic acetylcholine receptors
Kohei Nishimura et al.
ACS applied materials & interfaces, 11(34), 31105-31110 (2019-08-07)
In the composition of Q0.1(FA0.75MA0.25)0.9SnI3, Q is replaced with Na+, K+, Cs+, ethylammonium+ (EA+), and butylammonium+ (BA+), respectively, and the relationship between actually measured lattice strain and photovoltaic performances is discussed. The lattice strain evaluated by the Williamson-hall plot of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service