Merck
  • Diagnostic utility of immunohistochemistry in morphologically difficult prostate cancer: review of current literature.
Diagnostic utility of immunohistochemistry in morphologically difficult prostate cancer: review of current literature.
Histopathology (2005-06-29)
M Varma, B Jasani
ABSTRACT
Varma M & Jasani B (2005) Histopathology47, 1-16 Diagnostic utility of immunohistochemistry in morphologically difficult prostate cancer: review of current literatureImmunohistochemistry is widely used to distinguish prostate cancer from benign mimics and to establish the prostatic origin of poorly differentiated carcinoma. We critically review the recent advances in prostate cancer immunohistochemistry, including the introduction of newer basal cell markers such as p63 and the discovery of the overexpression of alpha-methylacyl coenzyme A racemase (AMACR) in prostate cancer. The description of newer urothelial markers to aid the distinction of prostate cancer from urothelial carcinoma is also presented together with refinements in the quality control of PSA and PSAP immunostaining. Although AMACR is a useful immunohistochemical marker for prostate cancer, it has significant limitations. These limitations are discussed and the need for interpreting AMACR immunoreactivity in the appropriate morphological context and in conjunction with basal call markers is emphasized. We also describe the utility of an immunohistochemical panel composed of PSA, PSAP and high molecular weight cytokeratin for distinguishing poorly differentiated prostate cancer from high-grade urothelial carcinoma. A morphological differential diagnosis based selection of immunohistochemical markers is highlighted as a novel approach in the diagnosis of prostate cancer in routine surgical pathology practice.
Sigma-Aldrich
PSA (EP109) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
PSA (ER-PR8) Mouse Monoclonal Antibody